Idex® Stainless Steel Sample Loop for Injection Valves
Square-cut, burr-free ends provide a firm, flush connection to valve ports
  • 316 stainless steel construction
  • High pressure resistance, low permeability
  • 30° port angle
A sample loop is a defined length of tubing and fittings configured to match the angle of the injection valve ports. A sample is loaded and held in the loop until injection is triggered, either automatically or manually. Stainless steel loops are supplied with unswaged fittings, so the tube can be completely bottomed in the injection port before the ferrule is swaged. The loop volumes are nominal.
$78.97 - $302.04CAD / Each
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
Compareitem specifications
Total Volume Capacity (µL) Tubing Inside Diameter (ID) (mm) Tubing Outside Diameter (OD) (in)
Availability Pricing
Idex Sample Loop for 7725/7725i Valves, SS, 20 µL, 1/16" OD x 0.012" ID; 1/EA 0201315 Mfr # 7755-022
COMPARE
20 0.3 0.0625
$78.97CAD / Each
Idex Sample Loop for 7725/7725i Valves, SS, 100 µL, 1/16" OD x 0.020" ID; 1/EA 0201316 Mfr # 7755-024
COMPARE
100 0.5 0.0625
$89.43CAD / Each
Idex Sample Loop for 7725/7725i Valves, SS, 5 µL, 1/16" OD x 0.007" ID; 1/EA 0201651 Mfr # 7755-020
COMPARE
5 0.18 0.0625
$89.43CAD / Each
Idex Sample Loop for 7725/7725i Valves, SS, 10 µL, 1/16" OD x 0.012" ID; 1/EA 0201652 Mfr # 7755-021
COMPARE
10 0.3 0.0625
$85.46CAD / Each
Idex Sample Loop for 7725/7725i Valves, SS, 50 µL, 1/16" OD x 0.020" ID; 1/EA 0201653 Mfr # 7755-023
COMPARE
50 0.5 0.0625
$85.75CAD / Each
Idex Sample Loop for 7725/7725i Valves, SS, 200 µL, 1/16" OD x 0.030" ID; 1/EA 0201654 Mfr # 7755-025
COMPARE
200 0.76 0.0625
$92.23CAD / Each
Idex Sample Loop for 7725/7725i Valves, SS, 500 µL, 1/16" OD x 0.030" ID; 1/EA 0201655 Mfr # 7755-026
COMPARE
500 0.76 0.0625
$103.73CAD / Each
Idex Sample Loop for 7725/7725i Valves, SS, 1 mL, 1/16" OD x 0.030" ID; 1/EA 0201656 Mfr # 7755-027
COMPARE
1000 0.76 0.0625
$120.23CAD / Each
Idex Sample Loop for 7725/7725i Valves, SS, 2 mL, 1/16" OD x 0.040" ID; 1/EA 0201657 Mfr # 7755-028
COMPARE
2000 1 0.0625
$175.48CAD / Each
Idex Sample Loop for 7725/7725i Valves, SS, 5 mL, 1/16" OD x 0.040" ID; 1/EA 0201658 Mfr # 7755-029
COMPARE
5000 1 0.0625
$302.04CAD / Each
Idex Sample Loop Kit for 7725/7725i Valves, SS, 1/16" OD; 1/EA 0201659 Mfr # 7755-060
COMPARE
200 0.0625
$117.28CAD / Each
Showing 1- 11 of 11