IDEX® PEEK Tubing, 100 Ft Length

An excellent replacement for stainless steel tubing
  • Flexible PEEK tubing is biocompatible and chemically inert
  • Easily cut to desired lengths
  • Smooth internal surface decreases turbulence
  • Color-coded for easier identification
Applications: Chromatography
Characteristics: Unlike stainless steel, PEEK (polyetheretherketone) tubing is flexible and easy to cut. It provides excellent chemical resistance to both organic and inorganic solvents. Tubing withstands pressures to 5000 psi (3000 psi for model 02006-30). PEEK tubing has a very smooth internal surface, which causes less turbulence than metal tubing and improves resolution. Pack of 5-ft.
Certification: None
Maximum temperature: 212°F (100°C)
Sterilize: With autoclave, ethylene oxide, or gamma irradiation
$505.08 - $1,331.94CAD / Each

13 variations of this product are available.

See All Product Options Below
13 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Tubing ID (in) Tubing OD (in)
Availability Pricing
Idex 1531BXL Chromatography Tubing, Blue PEEK, 1/16" OD x 0.010" ID x 100 ft L; 1/EA 0200621 Mfr # 1531BXL
COMPARE
Blue 1/100 1/16
$520.11CAD / Each
Idex 1531XL Chromatography Tubing, Natural PEEK, 1/16" OD x 0.010" ID x 100 ft L; 1/EA 0200624 Mfr # 1531XL
COMPARE
Natural 1/100 1/16
$520.11CAD / Each
Idex 1532XL Chromatography Tubing, Orange PEEK, 1/16" OD x 0.020" ID x 100 ft L; 1/EA 0200626 Mfr # 1532XL
COMPARE
Orange 1/50 1/16
$520.11CAD / Each
Idex 1533XL Chromatography Tubing, Green PEEK, 1/16" OD x 0.030" ID x 100 ft; 1/EA 0200629 Mfr # 1533XL
COMPARE
Green 1/32 1/16
$515.69CAD / Each
Idex 1535XL Chromatography Tubing, Red PEEK, 1/16" OD x 0.005" ID x 100 ft L; 1/EA 0200632 Mfr # 1535XL
COMPARE
Red 1/200 1/16
$731.54CAD / Each
Idex 1536XL Chromatography Tubing, Yellow PEEK, 1/16" OD x 0.007" ID x 100 ft L; 1/EA 0200634 Mfr # 1536XL
COMPARE
Yellow 1/150 1/16
$520.11CAD / Each
Idex 1538XL Chromatography Tubing, Natural PEEK, 1/16" OD x 0.040" ID x 100 ft L; 1/EA 0200637 Mfr # 1538XL
COMPARE
Natural 1/25 1/16
$505.08CAD / Each
Idex 1560XL Chromatography Tubing, Natural PEEK, 1/16" OD x 0.0025" ID x 100 ft L; 1/EA 0200639 Mfr # 1560XL
COMPARE
Natural 1/500 1/16
$748.48CAD / Each
Idex 1561XL Chromatography Tubing, Black PEEK, 1/16" OD x 0.004" ID x 100 ft L; 1/EA 0200641 Mfr # 1561XL
COMPARE
Black 1/250 1/16
$731.54CAD / Each
Idex 1568XL Chromatography Tubing, Natural PEEK, 1/32" OD x 0.015" ID x 100 ft L; 1/EA 0200643 Mfr # 1568XL
COMPARE
Natural 1/64 1/32
$874.46CAD / Each
Idex 1569XL Chromatography Tubing, Orange PEEK, 1/32" OD x 0.020" ID x 100 ft L; 1/EA 0200645 Mfr # 1569XL
COMPARE
Orange 1/50 1/32
$874.46CAD / Each
Idex 1576XL Chromatography Tubing, Red PEEK, 1/32" OD x 0.005" ID x 100 ft L; 1/EA 0200647 Mfr # 1576XL
COMPARE
Red 1/200 1/32
$1,331.94CAD / Each
Idex 1581XL Chromatography Tubing, Blue PEEK, 1/32" OD x 0.010" ID x 100 ft L; 1/EA 0200653 Mfr # 1581XL
COMPARE
Blue 1/100 1/32
$874.46CAD / Each
Showing 1- 13 of 13

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat