Idex® PEEK Sample Loops for Injection Valves
Biocompatible and inert to almost all organic solvents
  • Excellent alternative to stainless steel loops
  • Easy to install or replace
Biocompatible PEEK polymer sample loops are inert to almost all organic solvents, making them a versatile alternative to stainless steel. The loop ends are cut clean and straight for easy valve installation.
$245.17 - $368.35CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Material Compatible With Description
Availability Pricing
Idex Sample Loop for PR/EV700-100 & EV750-100, PEEK/Alumina Ceramic, 1 µL, Internal Groove; 1/EA 0201675 Mfr # 7123-227
COMPARE
PEEK/Alumina Ceramic Injection valve models PR/EV700-100 & EV750-100 Sample Loop for PR/EV700-100 & EV750-100, PEEK/Alumina Ceramic, 1 µL, Internal Groove; 1/EA
$368.35CAD / Each
Idex Sample Loop for 7725/7725i/9725/9725i/9010, PEEK/Alumina Ceramic, 2 µL, Internal Groove; 1/EA 0201676 Mfr # 7755-015
COMPARE
PEEK/Alumina Ceramic Injection valve 7725/7725i/9725/9725i/9010 Sample Loop for 7725/7725i/9725/9725i/9010, PEEK/Alumina Ceramic, 2 µL, Internal Groove; 1/EA
$245.17CAD / Each