Idex® Low-Pressure Unions with M6, 1/4-28, and 5/16-24 Flat-Bottom Configuration

Make solid connections in low-pressure applications
  • Manufactured from inert polymers
  • Provide your own fittings to suit your application
Use straight unions in a variety of low-pressure situations throughout your fluidic system. Use straight and elbow unions in a variety of low-pressure situations throughout your fluidic system. Unions do not include fittings, allowing for customization of fittings.
$8.40 - $24.75CAD / Each

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Fitting Material Description
Availability Pricing
Idex Low-Pressure Union, Male Straight, PEEK, 0.062" Bore, 1/4-28 Flat Bottom; 1/EA 0201407 Mfr # P-645
COMPARE
PEEK Low-Pressure Union, Male Straight, Natural PCTFE, 0.062" Bore, 1/4-28 Flat Bottom; 1/EA
$24.75CAD / Each
Idex Low-Pressure Union, Female Straight, Black Acetal, 0.020" Bore, M6 Flat Bottom; 1/EA 0202336 Mfr # P-602
COMPARE
Acetal Low-Pressure Union, Female Straight, Black Acetal, 0.020" Bore, M6 Flat Bottom; 1/EA
$8.40CAD / Each
Idex Low-Pressure Union, Female Straight, Blue ETFE, 0.020" Bore, M6 Flat Bottom; 1/EA 0202341 Mfr # P-622
COMPARE
ETFE Low-Pressure Union, Female Straight, Blue ETFE, 0.020" Bore, M6 Flat Bottom; 1/EA
$21.81CAD / Each
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat