Idex® Flangeless Standard Knurl Male Nuts, 1/4-28 Flat-Bottom Connection for 1/8" OD Tubing
Eliminate the need to flange tubing
  • Removable and reusable—saving you money!
Just slip the nut and ferrule over the tubing and finger tighten the assembly into your receiving port. Unlike the time required to flange tubing, it only takes a few seconds to change the components. Select from various ferrule materials to suit your application.
$2.21 - $55.69CAD / Each
26 variations of this product are available.
See All Product Options Below
26 PRODUCT OPTIONS
Compareitem specifications
Color Fitting Material Description
Availability Pricing
Idex Flangeless Male Nut, Standard Knurl, Natural PEEK, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 10/PK 0200766 Mfr # P-330X
COMPARE
Natural PEEK Flangeless Male Nut, 1/4-28 UNF for 1/8" OD Tubing, Peek, 10/Pk
$55.69CAD / Pkg of 10
Idex Flangeless Male Nut, Standard Knurl, Black Acetal, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA 0201984 Mfr # P-301
COMPARE
Black Acetal Flangeless Male Nut, Standard Knurl, Black Acetal, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
$2.36CAD / Each
Idex Flangeless Male Nut, Standard Knurl, Black Acetal, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 10/Pk 0201985 Mfr # P-301X
COMPARE
Black Acetal Flangeless Male Nut, Standard Knurl, Black Acetal, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 10/Pk
$23.43CAD / Pkg of 10
Idex Flangeless Male Nut, Standard Knurl, Red Acetal, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA 0201986 Mfr # P-302
COMPARE
Red Acetal Flangeless Male Nut, Standard Knurl, Red Acetal, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
$2.36CAD / Each
Idex Flangeless Male Nut, Standard Knurl, White Acetal, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA 0201987 Mfr # P-303
COMPARE
White Acetal Flangeless Male Nut, Standard Knurl, White Acetal, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
$2.36CAD / Each
Idex Flangeless Male Nut, Standard Knurl, Cream Acetal, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA 0201988 Mfr # P-304
COMPARE
Cream Acetal Flangeless Male Nut, Standard Knurl, Cream Acetal, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
$2.36CAD / Each
Idex Flangeless Male Nut, Standard Knurl, Green Acetal, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 10/PK 0201990 Mfr # P-305X
COMPARE
Green Acetal Flangeless Male Nut, Standard Knurl, Green Acetal, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 10/PK
$23.43CAD / Pkg of 10
Idex Flangeless Male Nut, Standard Knurl, Blue Acetal, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA 0201991 Mfr # P-306
COMPARE
Blue Acetal Flangeless Male Nut, Standard Knurl, Blue Acetal, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
$2.36CAD / Each
Idex Flangeless Male Nut, Standard Knurl, Natural ETFE, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA 0201998 Mfr # P-315
COMPARE
Natural ETFE Flangeless Male Nut, Standard Knurl, Natural ETFE, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
$4.57CAD / Each
Idex Flangeless Male Nut, Standard Knurl, Natural PP, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA 0202010 Mfr # P-320
COMPARE
Natural PP Flangeless Male Nut, Standard Knurl, Natural PP, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
$2.36CAD / Each
Idex Flangeless Male Nut, Standard Knurl, Natural PEEK, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/Ea 0202016 Mfr # P-330
COMPARE
Natural PEEK Flangeless Male Nut, Standard Knurl, Natural PEEK, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/Ea
$5.75CAD / Each
Idex Flangeless Male Nut, Standard Knurl, Blue PEEK, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 10/PK 0202017 Mfr # P-330B
COMPARE
Blue PEEK Flangeless Male Nut, Standard Knurl, Blue PEEK, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 10/PK
$6.04CAD / Each
Idex Flangeless Male Nut, Standard Knurl, Black PEEK, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA 0202018 Mfr # P-330BLK
COMPARE
Black PEEK Flangeless Male Nut, Standard Knurl, Black PEEK, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
$5.75CAD / Each
Idex Flangeless Male Nut, Standard Knurl, Green PEEK, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA 0202019 Mfr # P-330G
COMPARE
Green PEEK Flangeless Male Nut, Standard Knurl, Green PEEK, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
$6.04CAD / Each
Idex Flangeless Male Nut, Standard Knurl, Orange PEEK, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA 0202020 Mfr # P-330OR
COMPARE
Orange PEEK Flangeless Male Nut, Standard Knurl, Orange PEEK, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
$7.51CAD / Each
Idex Flangeless Male Nut, Standard Knurl, Red PEEK, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA 0202021 Mfr # P-330R
COMPARE
Red PEEK Flangeless Male Nut, Standard Knurl, Red PEEK, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
$11.05CAD / Each
Idex Flangeless Male Nut, Standard Knurl, Yellow PEEK, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA 0202022 Mfr # P-330Y
COMPARE
Yellow PEEK Flangeless Male Nut, Standard Knurl, Yellow PEEK, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
$5.75CAD / Each
Idex Flangeless Male Nut, Standard Knurl, Purple Acetal, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA 0202030 Mfr # P-338
COMPARE
Purple Acetal Flangeless Male Nut, Standard Knurl, Purple Acetal, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA
$2.36CAD / Each
Idex Flangeless Male Nut, Standard Knurl, Natural PFA, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA 0202037 Mfr # P-345
COMPARE
Natural PFA Flangeless Male Nut, Standard Knurl, Natural PFA, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
$3.83CAD / Each
Idex Flangeless Male Nut, Standard Knurl, Natural PFA, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 10/PK 0202038 Mfr # P-345X
COMPARE
Natural PFA Flangeless Male Nut, Standard Knurl, Natural PFA, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 10/PK
$39.63CAD / Pkg of 10
Showing 1 - 20 of 26