Idex® Flangeless Short Head Male Nuts, 1/4-28 Flat-Bottom Connection for 1/16" OD Tubing
Short head is ideal for tight-space applications
  • Eliminate the need to flange tubing
  • Removable and reusable—saving you money!
Just slip the nut and ferrule over the tubing and finger tighten the assembly into your receiving port. Unlike the time required to flange tubing, it only takes a few seconds to change the components. Select from various ferrule materials to suit your application.
$2.65 - $55.69CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Color Fitting Material Thread size UNF(M) (port 2) (InchShort)
Availability Pricing
Idex Flangeless Male Nut, Standard Head, Black Acetal, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 1/EA 0201947 Mfr # P-208
COMPARE
Black Acetal 1/4-28 flat bottom
$2.65CAD / Each
Idex Flangeless Male Nut, Standard Head, Natural ETFE, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 1/EA 0201956 Mfr # P-218
COMPARE
Natural ETFE 1/4-28 flat bottom
$3.83CAD / Each
Idex Flangeless Male Nut, Standard Head, Natural PEEK, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 1/EA 0201970 Mfr # P-235
COMPARE
Natural PEEK 1/4-28 flat bottom
$5.75CAD / Each
Idex Flangeless Male Nut, Standard Head, Natural PEEK, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 10/PK 0201972 Mfr # P-235X
COMPARE
Natural PEEK 1/4-28 flat bottom
$55.69CAD / Pkg of 10