Idex® Flangeless Large Knurl Fitting Assemblies, 1/2-20 Flat-Bottom Connection
Eliminate the need to flange tubing
  • Removable and reusable—saving you money!
Just slip the nut and ferrule over the tubing and finger tighten the assembly into your receiving port. Unlike the time required to flange tubing, it only takes a few seconds to change the components.
$14.00 - $127.74CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 1) (in) Description
Availability Pricing
Idex Flangeless Fitting, Large Knurl, Black PEEK, 1/4" OD Tubing, 1/2-20 Flat-Bottom; 1/Ea 0202103 Mfr # XU-655
COMPARE
1/4 Flangeless Fitting, Large Knurl, Black PEEK, 1/4" OD Tubing, 1/2-20 Flat-Bottom
$14.00CAD / Each
Idex Flangeless Fitting, Large Knurl, Black PEEK, 1/4" OD Tubing, 1/2-20 Flat-Bottom; 10/PK 0202104 Mfr # XU-655X
COMPARE
1/4 Flangeless Fitting, Large Knurl, Black PEEK, 1/4" OD Tubing, 1/2-20 Flat-Bottom;
$127.74CAD / Pkg of 10
Idex Flangeless Fitting, Large Knurl, Black PEEK, 5/16" OD Tubing, 1/2-20 Flat-Bottom; 1/EA 0202105 Mfr # XU-662
COMPARE
5/16 Flangeless Fitting, Large Knurl, Black PEEK, 5/16" OD Tubing, 1/2-20 Flat-Bottom; 1/EA
$15.91CAD / Each