Idex® Flanged Fittings, Standard Knurl Head with 1/4-28 Flat-Bottom Connection
Excellent for low to moderate pressure applications
  • Choice of fittings for 1/16" or 1/8" OD tubing
  • Multiple head styles and materials
  • Choice of colors
These chromatography fittings of nuts and washers are used with flanged tubing. Flanged nuts are available in Delrin® acetal to resist cross-threading or loosening while in use or choose ethylene tetrafluoroethylene (ETFE) for chemically aggressive environments. The nuts come with stainless steel washers.
$3.54 - $6.48CAD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Compression Size / Tube Od (Port 1) (in) Fitting Material
Availability Pricing
Idex Flanged Fitting, Standard Knurl, Blue Acetal, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 1/EA 0202563 Mfr # P-482B
COMPARE
Blue 1/16 Acetal
$4.27CAD / Each
Idex Flanged Fitting, Standard Knurl, Black Acetal, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 1/EA 0202564 Mfr # P-482BLK
COMPARE
Black 1/16 Acetal
$5.01CAD / Each
Idex Flanged Fitting, Standard Knurl, Natural ETFE, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 1/EA 0202567 Mfr # P-482T
COMPARE
Natural 1/16 ETFE
$3.54CAD / Each
Idex Flanged Fitting, Standard Knurl, Blue Acetal, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA 0202579 Mfr # P-582B
COMPARE
Blue 1/8 Acetal
$6.19CAD / Each
Idex Flanged Fitting, Standard Knurl, Natural ETFE, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA 0202580 Mfr # P-582T
COMPARE
Natural 1/8 ETFE
$6.48CAD / Each