Idex® Flanged Fittings, 5/16" Square Head with 1/4-28 Flat-Bottom Connection
Excellent for low to moderate pressure applications
  • Choice of fittings for 1/16" or 1/8" OD tubing
  • Multiple head styles and materials
  • Choice of colors
These chromatography fittings of nuts and washers are used with flanged tubing. Flanged nuts are available in Delrin® acetal to resist cross-threading or loosening while in use or choose ethylene tetrafluoroethylene (ETFE) for chemically aggressive environments. The nuts come with stainless steel washers.
$3.54 - $5.01CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Compression Size / Tube Od (Port 1) (in) Fitting Material
Availability Pricing
Idex Flanged Fitting, 5/16" Square, Black Acetal, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 1/EA 0202554 Mfr # P-401
COMPARE
Black 1/16 Acetal
$5.01CAD / Each
Idex Flanged Fitting, 5/16" Square, Black Acetal, 1/18" OD Tubing, 1/4-28 Flat-Bottom; 1/EA 0202569 Mfr # P-501
COMPARE
Black 1/8 Acetal
$3.54CAD / Each