Idex® Barbed Y Connectors
Make solid connections in low-pressure applications
  • Offers less turbulence and gentler mixing than Tee connectors
  • Manufactured from polypropylene
Use barbed Y connectors to firmly join together three pieces of soft-walled peristaltic tubing with similar inner diameters.
$2.80 - $6.48CAD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Barb Size / Tubing Id (Port 2) (in) Barb Size / Tubing Id (Port 3) (in)
Availability Pricing
Idex Barbed Connector, Y, Polypropylene, 1/16" Hose Barb; 1/EA 0202407 Mfr # P-860
COMPARE
1/16 1/16 1/16
$2.80CAD / Each
Idex Barbed Connector, Y, Polypropylene, 3/32" Hose Barb; 1/EA 0202408 Mfr # P-861
COMPARE
3/32 3/32 3/32
$4.27CAD / Each
Idex Barbed Connector, Y, Polypropylene, 1/8" Hose Barb; 1/EA 0202409 Mfr # P-862
COMPARE
1/8 1/8 1/8
$2.80CAD / Each
Idex Barbed Connector, Y, Polypropylene, 3/16" Hose Barb; 1/EA 0202410 Mfr # P-863
COMPARE
3/16 3/16 3/16
$6.48CAD / Each
Idex Barbed Connector, Y, Polypropylene, 1/4" Hose Barb; 1/EA 0202411 Mfr # P-864
COMPARE
1/4 1/4 1/4
$5.75CAD / Each