Idex® Barbed Luer Adapters
Excellent for connecting soft-wall tubing to a syringe or a low-pressure filter with luer connection
  • Choose from polypropylene or ETFE construction
Female luer fittings come with a lock. Male luer fittings are available with slip or lock connection.
ITEM(S) ON SALE
$1.62 - $5.10CAD / Each
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size-Port 2 (in) Connection Type-Port 1 Material
Availability Pricing
Idex Barbed Luer Adapter, Polypropylene, 0.030" Bore, Female Luer Lock x 1/16" Hose Barb; 1/EA 0201458 Mfr # P-857
COMPARE
1/16 Female luer lock Polypropylene
$1.62CAD / Each
Idex Barbed Luer Adapter, Polypropylene, 0.049" Bore, Male Luer Lock x 1/16" Hose Barb; 1/EA 0202398 Mfr # P-850
COMPARE
1/16 Male luer lock Polypropylene
$1.62CAD / Each
Idex Barbed Luer Adapter, Polypropylene, 0.071" Bore, Male Luer Lock x 3/32" Hose Barb; 1/EA 0202399 Mfr # P-851
COMPARE
3/32 Male luer lock Polypropylene
$1.62CAD / Each
Idex Barbed Luer Adapter, Polypropylene, 0.099" Bore, Male Luer Lock x 1/8" Hose Barb; 1/EA 0202400 Mfr # P-852
COMPARE
1/8 Male luer lock Polypropylene
$1.62CAD / Each
Idex Barbed Luer Adapter, Polypropylene, 0.108" Bore, Male Luer Lock x 5/32" Hose Barb; 1/EA 0202401 Mfr # P-853
COMPARE
5/32 Male luer lock Polypropylene
$2.80CAD / Each
Idex Barbed Luer Adapter, Polypropylene, 0.046" Bore, Male Slip Luer x 1/16" Hose Barb; 1/EA 0202402 Mfr # P-854
COMPARE
1/16 Male slip luer Polypropylene
$1.62CAD / Each
Idex Barbed Luer Adapter, Polypropylene, 0.069" Bore, Male Slip Luer x 3/32" Hose Barb; 1/EA 0202403 Mfr # P-855
COMPARE
3/32 Male slip luer Polypropylene
$1.62CAD / Each
Idex Barbed Luer Adapter, Polypropylene, 0.096" Bore Male Slip Luer x 1/8" Hose Barb; 1/EA 0202404 Mfr # P-856
COMPARE
1/8 Male slip luer Polypropylene
$2.80CAD / Each
Idex Barbed Luer Adapter, Polypropylene, 0.056" Bore, Female Luer Lock x 3/32" HoseBarb; 1/EA 0202405 Mfr # P-858
COMPARE
3/32 Female luer lock Polypropylene
$1.62CAD / Each
Idex Barbed Luer Adapter, Polypropylene, 0.080" Bore, Female Luer Lock x 1/8" Hose Barb; 1/EA 0202406 Mfr # P-859
COMPARE
1/8 Female luer lock Polypropylene
$1.62CAD / Each
Idex Barbed Luer Adapter, Natural ETFE, 0.030" Bore, Female Luer Lock x 1/16" Hose Barb; 1/EA 0202412 Mfr # P-870
COMPARE
1/16 Female luer lock ETFE
$5.10CAD / Each
$6.92CAD / Each
Showing 1- 11 of 11