Idex® Back Pressure Regulator (BPR) Cartridges
Select from preset pressure settings
  • Use with BPR holders
Use BPR cartridges with the holders to enhance system performance through outgassing prevention and improved pump check valve efficiency.
$133.49 - $145.13CAD / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Description Pressure Setting (PSI)
Availability Pricing
Idex Back Pressure Regulator Cartridge, 100 psi; 1/EA 0201441 Mfr # P-763
COMPARE
Idex P-763 Back Pressure Regulator Cartridge, 100 psi; 1/EA 100
$135.85CAD / Each
Idex Back Pressure Regulator Cartridge, 40 psi; 1/EA 0201714 Mfr # P-761
COMPARE
Idex Back Pressure Regulator Cartridge, 40 psi; 1/EA 40
$134.52CAD / Each
Idex Back Pressure Regulator Cartridge, 75 psi; 1/EA 0201716 Mfr # P-762
COMPARE
Idex Back Pressure Regulator Cartridge, 75 psi; 1/EA 75
$134.52CAD / Each
Idex Back Pressure Regulator Cartridge, 250 psi; 1/EA 0201717 Mfr # P-764
COMPARE
Idex Back Pressure Regulator Cartridge, 250 psi; 1/EA 250
$133.49CAD / Each
Idex Back Pressure Regulator Cartridge, 500 psi; 1/EA 0201718 Mfr # P-765
COMPARE
Idex Back Pressure Regulator Cartridge, 500 psi; 1/EA 500
$136.14CAD / Each
Idex Back Pressure Regulator Cartridge, 750 psi; 1/EA 0201726 Mfr # P-795
COMPARE
Idex Back Pressure Regulator Cartridge, 750 psi; 1/EA 750
$143.66CAD / Each
Idex Back Pressure Regulator Cartridge, 1000 psi; 1/EA 0201727 Mfr # P-796
COMPARE
Idex Back Pressure Regulator Cartridge, 1000 psi; 1/EA 1000
$145.13CAD / Each
Showing 1- 7 of 7