IC Biomedical LD Series Liquid Nitrogen Dewars
Secure, efficient, and built to deliver unmatched performance for safe and convenient liquid nitrogen handling
  • Durable ribbed aluminum body with magneformed necktube for maximum structural integrity
  • Advanced insulation ensures high thermal efficiency and longer holding times
  • Lightweight and easy to handle for lab or industrial environments
  • Resilient paint finish resists wear and corrosion
The LD Series LN2 Liquid Dewars are expertly engineered for the safe storage and controlled dispensing of small quantities of liquid nitrogen. Built with a rugged ribbed aluminum body and a magneformed necktube design, these containers offer exceptional strength and durability, even in demanding environments. Enhanced by advanced insulation materials, they provide superior thermal efficiency to minimize nitrogen loss. Whether in laboratory or industrial settings, the LD Series offers a reliable, user-friendly solution for all your cryogenic needs.
ITEM(S) ON SALE
$1,676.72 - $2,865.74CAD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Static Holding Time (days) Volume (Liters)
Availability Pricing
IC Biomedical 4LDB Liquid Nitrogen Storage Dewar, 4 L, 10 Day Static Hold 0377351 Mfr # 4LDB
COMPARE
10 4
$1,754.81CAD / Each
IC Biomedical 5LD Liquid Nitrogen Storage Dewar, 5 L, 6 Day Static Hold 0377353 Mfr # 5LD
COMPARE
6 5
$2,394.26CAD / Each
IC Biomedical 10LD Liquid Nitrogen Storage Dewar, 10 L, 45 Day Static Hold 0377355 Mfr # 10LD
COMPARE
45 10
$1,676.72CAD / Each
IC Biomedical 25LD Liquid Nitrogen Storage Dewar, 25 L, 109 Day Static Hold 0377357 Mfr # 25LD
COMPARE
109 25
Call For Price
IC Biomedical 35LD Liquid Nitrogen Storage Dewar, 35 L, 152 Day Static Hold 0377359 Mfr # 35LD
COMPARE
152 35
$2,865.74CAD / Each
IC Biomedical 50LD Liquid Nitrogen Storage Dewar, 50 L, 122 Day Static Hold 0377361 Mfr # 50LD
COMPARE
122 50
$2,764.85CAD / Each
$3,686.42CAD / Each