Horiba U-52G Multiparameter Meter
Measure and display pH, mV, conductivity, TDS, salinity, dissolved oxygen, turbidity, seawater specific gravity, water depth, and temperature simultaneously
  • Auto-calibration feature simplifies calibration of pH, dissolved oxygen, conductivity, turbidity, and depth
  • Built-in GPS — ideal for environmental surveys
  • Rugged housing is both waterproof and shock resistant
  • Stores up to 10,000 readings with USB connectivity for export of data to a PC
These handheld, waterproof meters feature a large 3-1/2" (8.9 cm), easy-to-read LCD with backlighting for use in dark and low-light conditions. Multiple parameters can be measured and displayed at the same time. USB/probe connectivity, GPS info, and battery power are also indicated via the display. Variable contrast allows you to adjust for extreme ambient light conditions.
Meters are designed for one-handed operation. Probes are attached via a quick-connect connector allowing for fast setup. Unique auto-calibration feature provides hassle-free meter setup. Turbidity sensor offers better accuracy than most other conventional water quality meters. Polarographic DO sensor features improved stability, fast response, and easy maintenance.
Auto-hold function freezes average data values providing more time to verify or transcribe data. Up to 10,000 data sets can be stored and later transferred to a PC via a built-in USB connection. Units of measure are user selectable which helps you save time.
$11,216.18 - $13,454.78CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Conductivity Max pH (pH) Description
Availability Pricing
Horiba 3200156563 Multiparameter Meter, 6-ft (2-m) Cable 9960025 Mfr # 3200156563
COMPARE
100 mS 14 Multiparameter Meter, 6-ft (2-m) Cable
$11,216.18CAD / Each
Horiba U-52G Multiparameter Meter, 100-ft (30-m) Cable 9960027 Mfr # 3200164500
COMPARE
100 mS 14 Multiparameter Meter, 100-ft (30-m) Cable
$13,454.78CAD / Each