Honeywell Weight-Balancing Arc Flash Protective Helmet and Face Shield
Unique weight-balancing design allows the face shield to sit in a balanced, compact position centered over the top of the hard hat when not needed. The shield easily slides into stowed position with one hand and can be quickly removed from the hard hat. Upper bracket ventilation features reduces fogging and lowers CO2 levels.
  • 12 cal/cm2 Arc Thermal Protective Value (ATPV) rating
  • 7-1/2" x 20" (191 x 508 mm) viewing area
  • Extra light tint for increased visibility
  • Absorbs >99.9% of harmful UV radiation
  • Meets ANSI Z87.1 and tested to ASTM F2178 specifications
  • Optional antifog inner lens features an air-spaced design that further decreases fogging
  • Optional task light features a high-intensity LED light; purchase two for lighting on both sides of the face shield
  • Optional transparent chin guard increases your peripheral view for improved awareness and safety
ITEM(S) ON SALE
$95.48 - $132.20CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Honeywell AFLENS Salisbury Optional Anti-Fog Inner Lens for Arc Flash Face Shield 8643617 Mfr # AFLENS-ES
COMPARE
Optional Anti-Fog Inner Lens for Arc Flash Face Shield
$95.48CAD / Each
Honeywell FLKIT Salisbury Optional Attachable Task Light with Clip for Arc Flash Face Shield 8643618 Mfr # FLKIT
COMPARE
Optional Attachable Task Light with Clip for Arc Flash Face Shield
$132.20CAD / Each
$197.43CAD / Each