Hirschmann® opus® Dispensers
Facilitates rapid aeration without loss of reagent
  • Electronic drive ensures repeatable results
  • Stores up to nine user-defined programs
  • USB and RS-232 interface provide direct PC connection
  • Compliant with DIN EN ISO 8655
The motorized liquid dispenser with a reagent recirculation system provides accurate and reliable serial dispensing results up to 9999 times. The dispensing procedure is configured on an external control module featuring a user-friendly IFT touch screen. All tasks are easily and quickly configured on the main screen; up to nine different user-defined programs can be saved in memory and reactivated at any time. The last setting is automatically saved when unit is turned off. Unique stepper function allows you to dispense several different volumes from a single cylinder filling. Recirculation system makes it possible for rapid venting when priming the pump without loss of reagent.
Universal luer connection lets you to add filters and drying tubes to the discharge path. Dispenser rotates freely on the bottle, providing flexibility. Quick-Cal function allows you to calibrate the dispenser for different temperatures and reagents. For added security, the PIN function protects against unauthorized access or can be configured to enable only selected functions. The ergonomically designed dispenser also features a clear inspection window which can be replaced with a colored UV protection window for light-sensitive media.
$6,563.95 - $7,035.44CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Accuracy Max Dispense Volume (mL) Min Dispense Volume (mL)
Availability Pricing
Hirschmann opus® Dispenser, 20 mL, 100 to 240 VAC 2550302 Mfr # 9581020
COMPARE
Dispenser: R=0.6%, CV=-0.2%; Stepper: R=0.5%, CV=0.3% 200 0.1
$6,563.95CAD / Each
Hirschmann opus® Dispenser, 50 mL, 100 to 240 VAC 2550303 Mfr # 9581050
COMPARE
Dispenser: R=0.6%, CV=-0.2%; Stepper: R=0.5%, CV=0.25% 50 1
$7,035.44CAD / Each