Heated Dolly for 55 Gallon Metal Drums
Mobile drum heater provides operator flexibility
  • Dolly supports up to 900 pound capacity
Use this drum heated dolly to warm your nonflammable, nonhazardous material contained within 55-gallon metal drums. The built-in infinite control (0 to 100% adjustable) features on/off as well as heat pulsing action. Drums rest on a spring-loaded 15 1/4” square by 1/4” thick aluminum heat platen, and rolls on 3"-diameter steel swivel casters that feature two-wheel locking capability. Measure the temperature of drum contents with an external temperature measurement device (not included) for more precise temperature control.
$1,878.57CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Drum Size Max Temperature (° F) Power (VAC)
Availability Pricing
Expo Engineering HDD-55/120V Heated Drum Dolly, Fits 55 gal, 550° F, 120 V 0950025 Mfr # HDD-55/120V
COMPARE
55 gallons 550 120
$1,878.57CAD / Each
Expo Engineering HDD-55/240V Heated Drum Dolly, Fits 55 gal, 550° F, 240 V 0950030 Mfr # HDD-55/240V
COMPARE
55 gallons 550 240
$1,878.57CAD / Each