Handheld Coating Thickness Gauges
Measures the thickness of coatings quickly and non-destructively using magnetic induction and Eddy current methods
These handheld coating thickness gauges have an integrated measurement probe for quick and easy results. The probe measures non-magnetic coatings like paint and enamel applied to ferrous substrates such as steel or iron using the magnetic induction method and/or measures non-conductive coatings applied to non-ferrous substrates such as aluminum and stainless steel, copper or brass, using the eddy current method. Gauges conform to ASTM D7091, B499, B244, DIN EN ISO 2178, 2360, 2808, SSPC-PA2 and more.
MPO Series features include display of mean value, standard deviation, min/max, number of measurements per block, and memory storage capability of up to 1000 measurements.
MPOR Series include a USB interface for bi-directional data transmission to Fischer Data Center Software allowing you download and store measurement data and create custom inspection reports. Gauges feature memory storage capability of up to 10,000 measurements.
$1,977.14 - $2,142.68CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Measuring Method Description
Availability Pricing
Fischer Technology MPO Dualscope Coating Thickness Gauge; 0-80 mils, Mag Induction/Eddy Current 5410219 Mfr # 605-360
COMPARE
Eddy current, Magnetic induction Coating Thickness Gauge; 0-80 mils, Mag Induction/Eddy Current
$1,977.14CAD / Each
Fischer Technology MPOR Dualscope Coating Thickness Gauge; 0-80 mils, Mag Induction/Eddy Current 5410222 Mfr # 605-097
COMPARE
Eddy current, Magnetic induction Coating Thickness Gauge; 0-80 mils, Mag Induction/Eddy Current
$2,142.68CAD / Each