H-B Instrument Digital Min/Max Indoor/Outdoor Thermometer
Simultaneously displays current, maximum, and minimum temperatures
  • Switch between indoor/outdoor temperature readings
  • Super-large LCD
The meter simultaneously displays current, maximum, and minimum temperatures with the ability to switch between indoor and outdoor temperature modes. Readings are shown on an extra large LCD that is easily viewed from several feet away, with the current temperature displayed by a bar scale. The unit features a Min/Max function that offers the highest and lowest readings from when the reset button was last pressed. Other features include a remote probe for outdoor readings, high/low alarm, low battery indicator, and ºF/°C selection.
$236.48CAD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Meter Measurement Type Number Of Probe Channels
Availability Pricing
H-B Instrument B60900-2800 DURAC Probeless Electronic Indoor/Outdoor Thermometer; -40/50C (-40/122F) 0808112 Mfr # B609002800
COMPARE
122 Digital Indicator
$236.48CAD / Each