Glass-Encased Stir Bars
  • Borosilicate glass-encased magnetic stir bars are completely non-reactive, leak proof, and nonabsorbent
  • Handles temperatures up to 525°F (274°C)
$33.74 - $63.95CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Diameter (in) Length (in)
Availability Pricing
COMPARE
0.375 1/2
$46.12CAD / Each
COMPARE
0.25 7/8
$49.21CAD / Each
COMPARE
0.375 1
$63.95CAD / Each
COMPARE
0.375 1 1/2
$33.74CAD / Each