Glas-Col Flexible Fabric Hemispherical Heating Mantles-240 VAC
Flexible fabric conforms to your vessel providing uniform heating
Hemispherical heating mantles feature insulated glass fabric that conforms to unusual vessel shapes. Steel extension supports (sold separately) are required to support mantles; do not place mantle directly on ring stand. Mantles require a controlled electrical power source to prevent overheating.
$495.35 - $767.78CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flask Size (mL) Power (VAC)
Availability Pricing
Glas-Col 100A O401 Flexible Fabric Heating Mantle, 200 ml, 100 Watts, 240 V 3622507 Mfr # 100A O401
COMPARE
200 240
$495.35CAD / Each
Glas-Col 100A O403 Flexible Fabric Heating Mantle, 250 ml, 180 Watts, 240 V 3622515 Mfr # 100A O403
COMPARE
250 240
$591.86CAD / Each
Glas-Col 100A O407 Flexible Fabric Heating Mantle, 500 ml, 270 Watts, 240 V 3622535 Mfr # 100A O407
COMPARE
500 240
$573.59CAD / Each
Glas-Col 100A O411 Flexible Fabric Heating Mantle, 2000 ml, 500 Watts, 240 V 3622555 Mfr # 100A O411
COMPARE
2000 240
$767.78CAD / Each