Fowler Ultra-Cal V Electronic Calipers
Choose from RS-232 or Bluetooth® output for data recording
  • Hardened stainless steel construction provides durability and corrosion resistance
  • Exceptional Swiss measurement quality with resolution to 0.0005 in (0.01 mm)
  • Measure four ways—inside, outside, depth, and step
  • Features include data hold, and absolute and relative measurement modes
$1,541.17 - $3,540.56CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max range (in) Max range (mm) Description
Availability Pricing
Fowler 54-100-167-0 Ultra-Cal V Electronic Caliper with Bluetooth; 6"/150mm 0992543 Mfr # 54-100-167-0
COMPARE
6 150 Ultra-Cal V Electronic Caliper with Bluetooth; 6"/150mm
$1,541.17CAD / Each
Fowler 54-100-168-0 Ultra-Cal V Electronic Caliper with Bluetooth; 8"/200mm 0992544 Mfr # 54-100-168-0
COMPARE
8 200 Ultra-Cal V Electronic Caliper with Bluetooth; 8"/200mm
$2,890.79CAD / Each
Fowler 54-100-169-0 Ultra-Cal V Electronic Caliper with Bluetooth; 12"/300mm 0992545 Mfr # 54-100-169-0
COMPARE
12 300 Ultra-Cal V Electronic Caliper with Bluetooth; 12"/300mm
$3,540.56CAD / Each