Foam-Tipped Glassware Brushes
Ergonomic design makes it easy to rotate brush
  • Tough, yet nonabrasive brushes
  • Dishwasher safe
Ergonomically designed wide, flat handles makes for easy rotating of brush. Excellent for all wide mouthed bottles, culture, volumetric graduated, filtering and many other kinds of long flasks, jugs and cylinders. Brushes are tough, yet non abrasive, and dishwasher safe. It works equally well on both plastic and glass bottles. Use with hot water. No detergents needed.
$25.49 - $34.48CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Handle Length (in) Length (in)
Availability Pricing
COMPARE
7 12
$25.49CAD / Each
COMPARE
13 20
$34.48CAD / Each