Fluke 971 Thermohygrometer
Ergonomically designed meter with datalogger
  • Calculates dew point and wet bulb
  • Use as part of regular HVAC maintenance
The Fluke 971 meter is built for field use with an impact resistant housing, rugged holster, convenient belt clip, and bright backlit display. Includes degF/degC selection, Min/Max reading, hold feature, averaging function, and 99 record datalogging capacity. A large LCD simultaneously displays RH and temperature readings, along with calculating dew point and wet bulb.
Available for sale in the United States and Canada only.
$792.68CAD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Battery Max RH Range (%) Max Temperature (° F)
Availability Pricing
Fluke 971 Thermohygrometer, 5 to 95% RH/, -4 to 140F 3785120 Mfr # FLUKE-971
COMPARE
4 AAA (included) 95 140
$792.68CAD / Each