Fluke 27 II and 28 II Rugged IP67 Industrial Multimeters

Available for sale in the United States and Canada only Waterproof and dustproof design for use in harsh environments
  • True RMS AC voltage and current for accurate measurements on nonlinear signals (28 II)
  • Auto and manual ranging provide maximum flexibility
  • Measures
  • Measure up to 1000 V ac and dc
  • Measure up to 10 A (20 A for 30 seconds)
  • 10,000 µF capacitance range
  • Frequency to 200 kHz
  • Extended AC voltage bandwidth – 15 Hz to 20 khz
  • Built in thermometer conveniently allows you to take temperatures readings without having to carry a separate instrument
  • Resistance, continuity and diode test
  • Min/Max and average recording to capture variations automatically
  • True RMS ac voltage and current (28 II)
  • Unique function for accurate voltage and frequency measurements on adjustable speed motor drives and other electrically noisy equipment (28 II)
  • Features
  • IP67 waterproof & dustproof protection, impervious to the harsh environments
  • Designed to withstand 3 meter drop
  • Hi resolution 20,000 count display mode (28 II)
  • True-rms ac voltage and current for accurate measurements on non linear signals (28 II)
  • Average responding voltage and current for measurements (27 II)
  • Easy to read display with large digits and a bright, two-level backlight
  • New backlit keypad buttons allows for easy visibility in low-lit areas
  • Large display digits and 2 level bright white backlight for increased visibility
  • Reversible holster for added protection when not in use
  • Withstands hazardous 8,000 volt spikes caused by load switching and faults on industrial circuits and complies with second edition IEC and ANSI electrical safety standards
  • Improved sleep mode for long battery life
  • Relative mode to remove test lead resistance from low ohms measurements
  • Auto and manual ranging for maximum flexibility
  • Optional magnetic hanger for easy setup and viewing while freeing your hands for other tasks
  • MSHA approved for hazardous areas
The new Fluke 27 II and 28 II digital multimeters define a new standard for operating in harsh conditions with the features and accuracy to troubleshoot most electrical problems. Both meters have IP67 (waterproof and dustproof) rating, MSHA approvals, extended operating temperature range of 5 to 131°F (-15 to +55°C), and have been designed and tested to withstand a 10-ft (3-m) drop. These multimeters are built to work in the toughest environments as they can withstand hazardous 8000 volt spikes caused by load switching and faults on industrial circuits; units comply with second edition IEC and ANSI electrical safety standards. Use the relative mode function to remove test lead resistance from low ohms measurements.

Model 20034-42 also features a built-in thermometer, peak min/max, high-resolution 20,000-count display mode, and unique function for accurate voltage and frequency measurements on adjustable-speed motor drives and other electrically noisy equipment.
2 PRODUCT OPTIONS
Compareitem specifications
DC Max (Volts) DC Voltage Accuracy True Rms
Availability Pricing
Fluke 27 II Rugged IP67 Industrial Multimeter 2003441 Mfr # FLUKE-27 II
COMPARE
1000
$1,149.23CAD / Each
Fluke 28 II Rugged IP67 Industrial TRMS Multimeter 2003442 Mfr # FLUKE-28 II
COMPARE
1000
$1,181.64CAD / Each
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat