FLIR® Si2 Industrial Acoustic Imaging Cameras
Increase safety while avoiding costly failures by quickly detecting compressed air and gas leaks, mechanical faults, and partial discharges
  • Superior sound detection with 124 microphones
  • Buil-in automatic frequency tuning and distance measurement
  • Machine learning capabilities for accurate measurements
  • 12 MP color camera, 8x digital zoom for excellent image clarity
  • Upload or transfer snapshots and videos for further analysis
  • Effortless one-hand operation
  • IP54-rated for protection against water and dust
The Si2-series acoustic imaging cameras offer an effective way to address the inspection requirements for manufacturing, electrical, and utility industries. With its superior performance, ability to identify issues up to 200 m (656 ft) away, and detection and measurement with increased sensitivity, it is a perfect tool for ensuring operational efficiency and safety.
Each acoustic imaging camera features 124 microphones that help pick up very quiet noises even from a long distance—ideal for inspecting high-voltage systems. The imaging cameras use machine learning to distinguish the other characteristics of the sound pattern created by leaks and partial discharge (PD) from the background, ensuring accurate measurements. The captured sound, transposed to a 12 MP digital camera and displayed on a 5" touchscreen LCD, visually pinpoints the leak source and mechanical faults.
Upload snapshots and videos to FLIR Acoustic Camera Viewer (a cloud service) using a wi-fi connection for instant access on your computer and mobile devices. Use the included USB memory stick when wi-fi is not available to transfer snapshots or video files*. Lightweight and ergonomically designed imaging cameras are easy to hold with one hand, so you can keep one hand free to operate the camera or to inspect components. Use the built-in LED to illuminate dimly lit areas. A long-life battery pack makes these cameras excellent for extended field use.
$29,245.32 - $40,650.83CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Gas Measured Range Description
Availability Pricing
FLIR Si2-LD Industrial Acoustic Imaging Camera for Pressurized Leak and Mechanical Fault Detections 3975916 Mfr # T912339
COMPARE
Compressed air, hydrogen, CO2, methane, natural gas, helium, argon, ammonia Bandwidth: 2 to 130 kHz Industrial Acoustic Imaging Camera for Pressurized Leak and Mechanical Fault Detections
$29,245.32CAD / Each
FLIR Si2-PD Industrial Acoustic Imaging Camera for Partial Discharge Detection 3975917 Mfr # T912305
COMPARE
None Bandwidth: 2 to 130 kHz Industrial Acoustic Imaging Camera for Partial Discharge Detection
$30,779.12CAD / Each
FLIR Si2-Pro Industrial Acoustic Imaging Camera for Partial Discharge, Pressurized Leak, and Mechanical Fault Detections 3975918 Mfr # T912340
COMPARE
Compressed air, hydrogen, CO2, methane, natural gas, helium, argon, ammonia Bandwidth: 2 to 130 kHz Industrial Acoustic Imaging Camera for Partial Discharge, Pressurized Leak, and Mechanical Fault Detections
$40,650.83CAD / Each