Elmi Fugamix™ Microcentrifuge/Mixers
Mix and spin down in a single machine!
  • Automated spin-mix-spin program for sample preparation
  • Digital timer helps achieve consistent results
  • 2 year warranty
These unique microcentrifuges let you mix and spin in a single unit—ideal for biochemistry, molecular, cell biology, and immunology applications as well as clinical and PCR diagnostic labs. Select from multiple mixing levels depending on your sample. Compact design takes up little benchtop space.
$1,029.90 - $1,685.56CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max RCF x g Max Speed (rpm) Description
Availability Pricing
Elmi CM-70M-09 Fugamix™ Microcentrifuge/Mixer, 9000 rpm, 24V RK-85230-30 Mfr # CM-MC-10
COMPARE
5200 9000 Fugamix™ Microcentrifuge/Mixer, 9000 RPM, 24V
$1,029.90CAD / Each
Elmi CM-50M Advanced Fugamix™ Microcentrifuge/Mixer, 15,000 rpm, 24V RK-85230-38 Mfr # CM-MC-2
COMPARE
15294 15000 Advanced Fugamix™ Microcentrifuge/Mixer, 15,000 RPM, 24V
$1,685.56CAD / Each