Cole-Parmer® EH-200 Series Macro-Kjeldahl Extraction Heaters
Designed to determine the protein content of food on a macro scale—from 100 to 800 mL
  • Independent heat controller lets you adjust the temperature range of each mantle individually
  • Stainless steel construction is rugged and easy to clean
Run several projects simultaneously using these multiposition macro-kjeldahl extraction heaters. Heaters are available with two or six individually controlled heating mantles that accommodate either 100 to 300 mL vessels or 500 to 800 mL vessels. Back-mounted brackets hold flask support rods up to 12.7 mm in diameter.
Each mantle has an independent heater with control dial, on/off switch, and indicator light, and operates to a maximum element temperature of 800°C. A quartz heating element is housed in a thermally insulated cartridge for easy replacement, extending the life of the heater. Heat controllers located in the lower part of the unit is safely separated from the upper heating elements by a stainless steel tray and well-ventilated air space. Extra safety features include an earth screen and double fuses on the supply line.
$4,673.59 - $10,094.19CAD / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flask Size (mL) Number of Positions Description
Availability Pricing
COMPARE
300 2 2-Well Macro-Kjeldahl Extraction Heater, 300 mL, US; 115 VAC
$4,675.07CAD / Each
COMPARE
300 2 2-Well Macro-Kjeldahl Extraction Heater, 300 mL, UK; 230 VAC
$4,828.30CAD / Each
COMPARE
300 2 2-Well Macro-Kjeldahl Extraction Heater, 300 mL, EU; 230 VAC
$4,673.59CAD / Each
COMPARE
800 2 2-Well Macro-Kjeldahl Extraction Heater, 800 mL, US; 115 VAC
$5,376.40CAD / Each
COMPARE
800 2 2-Well Macro-Kjeldahl Extraction Heater, 800 mL, UK; 230 VAC
$5,227.59CAD / Each
COMPARE
800 2 2-Well Macro-Kjeldahl Extraction Heater, 800 mL, EU; 230 VAC
$5,058.15CAD / Each
COMPARE
300 6 6-well Macro-Kjeldahl Extraction Heater, 300 mL, US; 115 VAC
$8,494.09CAD / Each
COMPARE
300 6 6-well Macro-Kjeldahl Extraction Heater, 300 mL, UK; 230 VAC
$9,370.76CAD / Each
COMPARE
300 6 6-well Macro-Kjeldahl Extraction Heater, 300 mL, EU; 230 VAC
$8,402.74CAD / Each
COMPARE
800 6 6-well Macro-Kjeldahl Extraction Heater, 800 mL, US; 115 VAC
$8,648.80CAD / Each
COMPARE
800 6 6-well Macro-Kjeldahl Extraction Heater, 800 mL, EU; 230 VAC
$10,094.19CAD / Each
COMPARE
800 6 6-well Macro-Kjeldahl Extraction Heater, 800 mL, UK; 230 VAC
$9,139.44CAD / Each
Showing 1- 12 of 12