Eldex Optos Model 3 High-Pressure Piston Metering Pumps

Compact pumps provide precise flow rate control and smooth fluid delivery at high pressures
  • Reciprocating piston design for precise flow control
  • High-resolution stepper motor minimizes pulsation
  • Control flow remotely via analog or RS-232 connection
  • Integrated piston wash extends piston seal life
  • Solvent-resistant membrane keypad
  • Use with liquids with temperatures ranging from 5 to 260°C (41 to 500°F)
  • Compact meter only measures 10 cm W x 24 cm D (4” x 9.5”), saving valuable benchtop space
The Optos series metering pumps deliver high precision and reliability at high pressures. Achieve flow rates from 0.01 to 80 mL/min with ±0.3% reproducibility with Model 3 meters. The reciprocating piston offers precise flow control while an advanced software algorithms control high-resolution stepper motor to maximize flow output and minimize pulsation. Easily set flow rates using the up and down buttons on the solvent-resistant keypad. Remotely control your meter via an analog signal (4 to 20 mA or 0 to 5 VDC) or RS-232. Contact closures allow remote start and stop, flow output (0 to 5 VDC), and pump error output.

Select a pump that uses the upper half of the flow range listed for each pump. Piston cylinder comes available in two materials—316 stainless steel (SS) and PEEK. 316 SS offers excellent chemical and corrosion resistance; use PEEK for extremely corrosive fluids. A fully integrated piston wash system cleans out buffers or solutions which can crystallize and lead to abrasion of the piston seal, shortening the piston seal life.
$5,186.33 - $6,313.48CAD / Each

6 variations of this product are available.

See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (mL/min) Max Pressure (PSI) Wetted Materials
Availability Pricing
Eldex Optos Model 3LM Piston Metering Pump, 316 SS, 3/32" Piston Dia, 0.01 to 10 mL/min 4853182 Mfr # 5980
COMPARE
10 3000 316 stainless steel, sapphire, ruby, graphite fiber reinforced PTFE, PCTFE
$5,341.04CAD / Each
Eldex Optos Model 3SM Piston Metering Pump, 316 SS, 1/8" Piston Dia, 0.01 to 20 mL/min 4853183 Mfr # 5982
COMPARE
20 1500 316 stainless steel, sapphire, ruby, graphite fiber reinforced PTFE, PCTFE
$5,186.33CAD / Each
Eldex Optos Model 3HM Piston Metering Pump, 316 SS, 1/4" Piston Dia, 0.04 to 80 mL/min 4853184 Mfr # 5986
COMPARE
80 750 316 stainless steel, sapphire, ruby, graphite fiber reinforced PTFE, PCTFE
$5,488.38CAD / Each
Eldex Optos Model 3LI Piston Metering Pump, PEEK, 3/32" Piston Dia, 0.01 to 10 mL/min 4853188 Mfr # 5981
COMPARE
10 3000 PEEK, sapphire, ruby, UHMW polyethylene, PCTFE
$6,011.43CAD / Each
Optos Model 3SI Piston Metering Pump, PEEK, 1/8" Piston Dia, 0.01 to 20 mL/min 4853189 Mfr # 5983
COMPARE
20 1500 PEEK, sapphire, ruby, UHMW polyethylene, PCTFE
$6,011.43CAD / Each
Eldex Optos Model 3HI Piston Metering Pump, PEEK, 1/4" Piston Dia, 0.04 to 80 mL/min 4853190 Mfr # 5984
COMPARE
80 750 PEEK, sapphire, ruby, UHMW polyethylene, PCTFE
$6,313.48CAD / Each
Showing 1- 6 of 6

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat