Eldex Optos Model 2 High-Pressure Piston Metering Pumps

Compact pumps provide precise flow rate control and smooth fluid delivery at high pressures
  • Reciprocating piston design for precise flow control
  • High-resolution stepper motor minimizes pulsation
  • Control flow remotely via analog or RS-232 connection
  • Integrated piston wash extends piston seal life
  • Solvent-resistant membrane keypad
  • Use with liquids with temperatures ranging from 5 to 260°C (41 to 500°F)
  • Compact meter only measures 10 cm W x 24 cm D (4” x 9.5”), saving valuable benchtop space
The Optos series metering pumps deliver high precision and reliability at high pressures. Achieve flow rates from 0.003 to 40 mL/min with ±0.3% reproducibility with Model 2 meters. The reciprocating piston offers precise flow control while an advanced software algorithms control high-resolution stepper motor to maximize flow output and minimize pulsation. Easily set flow rates using the up and down buttons on the solvent-resistant keypad. Remotely control your meter via an analog signal (4 to 20 mA or 0 to 5 VDC) or RS-232. Contact closures allow remote start and stop, flow output (0 to 5 VDC), and pump error output.

Select a pump that uses the upper half of the flow range listed for each pump. Piston cylinder comes available in two materials—316 stainless steel (SS) and PEEK. 316 SS offers excellent chemical and corrosion resistance; use PEEK for extremely corrosive fluids. A fully integrated piston wash system cleans out buffers or solutions which can crystallize and lead to abrasion of the piston seal, shortening the piston seal life.
$5,186.33 - $6,313.48CAD / Each

6 variations of this product are available.

See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (mL/min) Max Pressure (PSI) Wetted Materials
Availability Pricing
Eldex Optos Model 2LM Piston Metering Pump, 316 SS, 3/32" Piston Dia, 0.003 to 5 mL/min 4853171 Mfr # 5972
COMPARE
5 6000 316 stainless steel, sapphire, ruby, graphite fiber reinforced PTFE, PCTFE
$5,341.04CAD / Each
Eldex Optos Model 2SM Piston Metering Pump, 316 SS, 1/8" Piston Dia, 0.01 to 10 mL/min 4853172 Mfr # 5976
COMPARE
10 6000 316 stainless steel, sapphire, ruby, graphite fiber reinforced PTFE, PCTFE
$5,186.33CAD / Each
Eldex Optos Model 2HM Piston Metering Pump, 316 SS, 1/4" Piston Dia, 0.02 to 40 mL/min 4853173 Mfr # 5979
COMPARE
40 1500 316 stainless steel, sapphire, ruby, graphite fiber reinforced PTFE, PCTFE
$5,657.82CAD / Each
Eldex Optos Model 2LI Piston Metering Pump, PEEK, 3/32" Piston Dia, 0.003 to 5 mL/min 4853177 Mfr # 5974
COMPARE
5 4000 PEEK, sapphire, ruby, UHMW polyethylene, PCTFE
$6,011.43CAD / Each
Eldex Optos Model 2SI Piston Metering Pump, PEEK, 1/8" Piston Dia, 0.01 to 10 mL/min 4853178 Mfr # 5977
COMPARE
10 4000 PEEK, sapphire, ruby, UHMW polyethylene, PCTFE
$6,011.43CAD / Each
Eldex Optos Model 2HI Piston Metering Pump, PEEK, 1/4" Piston Dia, 0.02 to 40 mL/min 4853179 Mfr # 5948
COMPARE
40 1500 PEEK, sapphire, ruby, UHMW polyethylene, PCTFE
$6,313.48CAD / Each
Showing 1- 6 of 6

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat