DuPont™ Liveo™ Pharma TPE Tubing
Improves heat-welding and high tensile strength
  • High resiliency
  • Provides excellent flexibility
  • Offers low extractables
  • Contains no peroxide by-products, chlorophenyls or PCBs
  • Free from phthalates or latex additives
  • Chemical resistance
  • Easily sterilized
  • Meets USP Class VI, ISO 10993 (Sections 5, 6, 11, and 23), and U.S. Pharmacopeia requirements
The translucent DuPont Liveo Pharma TPE tubing is ideal for use in many biopharma applications, including fluid, media and solvent transport; single-use assemblies; peristaltic pump applications; and aseptic connection and disconnection without connectors. It offers improved heat-welding and high tensile strength, as well as burst resistance before and after welding, The tubing features minimal spallation after 24 hours of pumping, stable clarity after sterilization, complete traceability, and consistent performance.
Made from medical grade elastomer, the tubing complies with United States Pharmacopeia (USP®) Class VI Plastics test requirements. It is manufactured to the principles of cGMPs for Pharmaceutical products and ISO 7 Class clean room. The tubing is easily sterilized by ethylene oxide, autoclave steam for 30 minutes at 120 °C, and gamma radiation at 50 kGy.
$181.96 - $779.42CAD / Pkg of 1
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (in) Tubing OD (in) Wall Thickness (in)
Availability Pricing
DuPont Liveo Pharma TPE Tubing, 1/8" ID x 1/4" OD; 50 Ft 5012230 Mfr # 12078352
COMPARE
1/8 1/4 1/16
$181.96CAD / Pkg of 1
DuPont Liveo Pharma TPE Tubing, 3/16" ID x 5/16" OD; 50 Ft 5012231 Mfr # 12089526
COMPARE
3/16 5/16 1/16
$196.70CAD / Pkg of 1
DuPont Liveo Pharma TPE Tubing, 1/4" ID x 1/2" OD; 50 Ft 5012232 Mfr # 12098419
COMPARE
1/4 1/2 1/8
$282.52CAD / Pkg of 1
DuPont Liveo Pharma TPE Tubing, 1/4" ID x 3/8" OD; 50 Ft 5012233 Mfr # 12089205
COMPARE
1/4 3/8 1/16
$222.11CAD / Pkg of 1
DuPont Liveo Pharma TPE Tubing, 1/4" ID x 7/16" OD; 50 Ft 5012234 Mfr # 12084560
COMPARE
1/4 7/16 3/32
$258.95CAD / Pkg of 1
DuPont Liveo Pharma TPE Tubing, 3/8" ID x 5/8" OD; 50 Ft 5012235 Mfr # 12078431
COMPARE
3/8 5/8 1/8
$369.08CAD / Pkg of 1
DuPont Liveo Pharma TPE Tubing, 3/8" ID x 9/16" OD; 50 Ft 5012236 Mfr # 12084559
COMPARE
3/8 9/16 3/32
$331.88CAD / Pkg of 1
DuPont Liveo Pharma TPE Tubing, 1/2" ID x 11/16" OD; 50 Ft 5012237 Mfr # 12089906
COMPARE
1/2 11/16 3/32
$464.12CAD / Pkg of 1
DuPont Liveo Pharma TPE Tubing, 1/2" ID x 3/4" OD; 50 Ft 5012238 Mfr # 12078432
COMPARE
1/2 3/4 1/8
$508.32CAD / Pkg of 1
DuPont Liveo Pharma TPE Tubing, 3/4" ID x 1" OD; 50 Ft 5012239 Mfr # 12089295
COMPARE
3/4 1 1/8
$779.42CAD / Pkg of 1
Showing 1- 10 of 10