DuPont™ Liveo™ Pharma Advanced Pump Platinum-Cured Silicone Tubing
Offers four times the pump life of standard platinum-cure silicone tubing
  • Superior resiliency
  • Provides excellent flexibility
  • Offers low extractables
  • Contains no peroxide by-products, chlorophenyls or PCBs
  • Free from organic plasticizers, phthalates or latex additives
  • Easily sterilized
  • Meets USP Class VI, FDA 21 CFR, and ISO 10993 (Sections 3, 4, 5, 6, 10, and 11) requirements
The translucent DuPont Liveo Pharma Advanced Pump Tubing (APT) is specifically designed for pumping ultra-pure liquids in pharmaceutical and biotechnological processes where contamination is a concern. It offers a non-wetting surface, outstanding filling accuracy, complete traceability, and consistent performance.
Made from biomedical grade tear resistant platinum-cured elastomer, the tubing allows you up to four times the pump life of standard platinum-cure tubing and exceeds United States Pharmacopeia (USP®) Class VI Plastics Test requirements. It also meets the European Pharmacopoeia monograph 3.1.9. “Silicone elastomer for closures and tubing.” The tubing is easily sterilized by ethylene oxide, autoclave steam for 30 minutes at 120 °C, and gamma radiation at 45 kGy.
ITEM(S) ON SALE
$185.80 - $901.45CAD / Pkg of 1
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (in) Tubing OD (in) Wall Thickness (in)
Availability Pricing
DuPont Liveo Pharma Advanced Pump Tubing, 1/8" ID x 1/4" OD; 50 Ft 5012217 Mfr # 4040492
COMPARE
1/8 1/4 1/16
$351.77CAD / Pkg of 1
DuPont Liveo Pharma Advanced Pump Tubing, 3/16" ID x 3/8" OD; 50 Ft 5012218 Mfr # 4076620
COMPARE
3/16 3/8 3/32
$288.00CAD / Pkg of 1
$629.87CAD / Pkg of 1
DuPont Liveo Pharma Advanced Pump Tubing, 3/16" ID x 5/16" OD; 50 Ft 5012219 Mfr # 4077165
COMPARE
3/16 5/16 1/16
$185.80CAD / Pkg of 1
$406.29CAD / Pkg of 1
DuPont Liveo Pharma Advanced Pump Tubing, 1/4" ID x 1/2" OD; 50 Ft 5012220 Mfr # 4040491
COMPARE
1/4 1/2 1/8
$416.20CAD / Pkg of 1
$900.24CAD / Pkg of 1
DuPont Liveo Pharma Advanced Pump Tubing, 1/4" ID x 3/8" OD; 50 Ft 5012221 Mfr # 4076619
COMPARE
1/4 3/8 1/16
$226.10CAD / Pkg of 1
$493.59CAD / Pkg of 1
DuPont Liveo Pharma Advanced Pump Tubing, 1/4" ID x 7/16" OD; 50 Ft 5012222 Mfr # 4076981
COMPARE
1/4 7/16 3/32
$285.15CAD / Pkg of 1
$621.77CAD / Pkg of 1
DuPont Liveo Pharma Advanced Pump Tubing, 3/8" ID x 1/2" OD; 50 Ft 5012223 Mfr # 4085723
COMPARE
3/8 1/2 1/16
$273.60CAD / Pkg of 1
$597.46CAD / Pkg of 1
DuPont Liveo Pharma Advanced Pump Tubing, 3/8" ID x 5/8" OD; 50 Ft 5012224 Mfr # 4040286
COMPARE
3/8 5/8 1/8
$568.80CAD / Pkg of 1
$1,230.28CAD / Pkg of 1
DuPont Liveo Pharma Advanced Pump Tubing, 3/8" ID x 9/16" OD; 50 Ft 5012225 Mfr # 4090087
COMPARE
3/8 9/16 3/32
$728.59CAD / Pkg of 1
DuPont Liveo Pharma Advanced Pump Tubing, 1/2" ID x 3/4" OD; 50 Ft 5012226 Mfr # 4040490
COMPARE
1/2 3/4 1/8
$620.65CAD / Pkg of 1
$1,343.73CAD / Pkg of 1
DuPont Liveo Pharma Advanced Pump Tubing, 1/2" ID x 11/16" OD; 50 Ft 5012227 Mfr # 4090085
COMPARE
1/2 11/16 3/32
$450.75CAD / Pkg of 1
$975.38CAD / Pkg of 1
DuPont Liveo Pharma Advanced Pump Tubing, 3/4" ID x 1" OD; 50 Ft 5012228 Mfr # 4090086
COMPARE
3/4 1 1/8
$901.45CAD / Pkg of 1
$1,937.51CAD / Pkg of 1
DuPont Liveo Pharma Advanced Pump Tubing, 5/8" ID x 7/8" OD; 50 Ft 5012229 Mfr # 4038450
COMPARE
5/8 7/8 1/8
$563.05CAD / Pkg of 1
$1,218.49CAD / Pkg of 1
Showing 1- 13 of 13