DuPont ™ Liveo™ Pharma-80 Platinum-Cured Silicone Tubing
Ideal for transfer of high-purity fluids in pharmaceutical and biotechnological applications
  • Highest kink resistance
  • Provides excellent flexibility
  • Offers low extractables
  • Contains no peroxide by-products, chlorophenyls or PCBs
  • Free from organic plasticizers, phthalates or latex additives
  • Easily sterilized
  • Meets USP Class VI, FDA 21 CFR, and ISO 10993 (Sections 3, 4, 5, 6, 10, and 11) requirements
The translucent, platinum-cured silicone DuPont Liveo Pharma-80 tubing is designed to transfer ultra-pure liquids in pharmaceutical and biotechnological processes where contamination is a concern. The tubing offers the highest kink resistance, is stable over a wide temperature range, is highly resilient, impairs no taste or odor, and offers a non-wetting surface.
Made from BioMedical Grade elastomer, the tubing exceeds United States Pharmacopeia (USP®) Class VI Plastics Test Requirements. It also meets the European Pharmacopoeia monograph 3.1.9. “Silicone elastomer for closures and tubing.” The tubing is easily sterilized by ethylene oxide, autoclave steam for 30 minutes at 120 °C, and gamma radiation at 50 kGy.
ITEM(S) ON SALE
$306.75 - $1,177.95CAD / Pkg of 1
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (in) Tubing OD (in) Wall Thickness (in)
Availability Pricing
DuPont Liveo Pharma-80 Platinum-Cured Silicone Tubing, 1/8" ID x 1/4" OD; 50 Ft 5012207 Mfr # 4090082
COMPARE
1/8 1/4 1/16
$458.96CAD / Pkg of 1
DuPont Liveo Pharma-80 Platinum-Cured Silicone Tubing, 3/16" ID x 5/16" OD; 50 Ft 5012208 Mfr # 4090084
COMPARE
3/16 5/16 1/16
$581.99CAD / Pkg of 1
DuPont Liveo Pharma-80 Platinum-Cured Silicone Tubing, 1/4" ID x 1/2" OD; 50 Ft 5012209 Mfr # 4008432
COMPARE
1/4 1/2 1/8
$1,155.14CAD / Pkg of 1
DuPont Liveo Pharma-80 Platinum-Cured Silicone Tubing, 1/4" ID x 3/8" OD; 50 Ft 5012210 Mfr # 4066764
COMPARE
1/4 3/8 1/16
$306.75CAD / Pkg of 1
$668.92CAD / Pkg of 1
DuPont Liveo Pharma-80 Platinum-Cured Silicone Tubing, 3/8" ID x 1/2" OD; 50 Ft 5012211 Mfr # 4079457
COMPARE
3/8 1/2 1/16
$364.35CAD / Pkg of 1
$786.79CAD / Pkg of 1
DuPont Liveo Pharma-80 Platinum-Cured Silicone Tubing, 3/8" ID x 5/8" OD; 50 Ft 5012212 Mfr # 3100651
COMPARE
3/8 5/8 1/8
$791.30CAD / Pkg of 1
$1,517.59CAD / Pkg of 1
DuPont Liveo Pharma-80 Platinum-Cured Silicone Tubing, 3/8" ID x 9/16" OD; 50 Ft 5012213 Mfr # 4079507
COMPARE
3/8 9/16 3/32
$486.75CAD / Pkg of 1
$1,052.00CAD / Pkg of 1
DuPont Liveo Pharma-80 Platinum-Cured Silicone Tubing, 1/2" ID x 3/4" OD; 50 Ft 5012214 Mfr # 4008433
COMPARE
1/2 3/4 1/8
$832.35CAD / Pkg of 1
$1,782.80CAD / Pkg of 1
DuPont Liveo Pharma-80 Platinum-Cured Silicone Tubing, 3/4" ID x 1" OD; 50 Ft 5012215 Mfr # 4008434
COMPARE
3/4 1 1/8
$1,177.95CAD / Pkg of 1
$2,526.86CAD / Pkg of 1
DuPont Liveo Pharma-80 Platinum-Cured Silicone Tubing, 5/8" ID x 7/8" OD; 50 Ft 5012216 Mfr # 4016844
COMPARE
5/8 7/8 1/8
$992.20CAD / Pkg of 1
$2,129.05CAD / Pkg of 1
Showing 1- 10 of 10