DuPont™ Liveo™ Pharma-65 Platinum-Cured Silicone Tubing
Ideal for transfer of high-purity fluids in pharmaceutical and biotechnological applications
  • Greater pressure and kink resistance
  • Provides excellent flexibility
  • Offers low extractables
  • Contains no peroxide by-products, chlorophenyls or PCBs
  • Free from organic plasticizers, phthalates or latex additives
  • Easily sterilized
  • Meets USP Class VI, FDA 21 CFR, and ISO 10993 (Sections 3, 4, 5, 6, 10, and 11) requirements
The translucent, platinum-cured silicone DuPont Liveo Pharma-65 tubing is designed to transfer ultra-pure liquids in pharmaceutical and biotechnological processes where contamination is a concern. The tubing offers greater pressure and kink resistance, is stable over a wide temperature range, is highly resilient, impairs no taste or odor, and offers a non-wetting surface.
Made from BioMedical Grade elastomer, the tubing exceeds United States Pharmacopeia (USP®) Class VI Plastics Test Requirements. It also meets the European Pharmacopoeia monograph 3.1.9. “Silicone elastomer for closures and tubing.” The tubing is easily sterilized by ethylene oxide, autoclave steam for 30 minutes at 120 °C, and gamma radiation at 50 kGy.
ITEM(S) ON SALE
$177.15 - $679.70CAD / Pkg of 1
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (in) Tubing OD (in) Wall Thickness (in)
Availability Pricing
DuPont Liveo Pharma-65 Platinum-Cured Silicone Tubing, 1/8" ID x 1/4" OD; 50 Ft 5012198 Mfr # 4090602
COMPARE
1/8 1/4 1/16
$275.52CAD / Pkg of 1
DuPont Liveo Pharma-65 Platinum-Cured Silicone Tubing, 3/16" ID x 3/8" OD; 50 Ft 5012199 Mfr # 4077179
COMPARE
3/16 3/8 3/32
$232.80CAD / Pkg of 1
$496.53CAD / Pkg of 1
DuPont Liveo Pharma-65 Platinum-Cured Silicone Tubing, 3/16" ID x 5/16" OD; 50 Ft 5012200 Mfr # 4041047
COMPARE
3/16 5/16 1/16
$419.05CAD / Pkg of 1
$907.61CAD / Pkg of 1
DuPont Liveo Pharma-65 Platinum-Cured Silicone Tubing, 1/4" ID x 1/2" OD; 50 Ft 5012201 Mfr # 4008344
COMPARE
1/4 1/2 1/8
$303.85CAD / Pkg of 1
$662.29CAD / Pkg of 1
DuPont Liveo Pharma-65 Platinum-Cured Silicone Tubing, 1/4" ID x 3/8" OD; 50 Ft 5012202 Mfr # 4077978
COMPARE
1/4 3/8 1/16
$177.15CAD / Pkg of 1
$386.03CAD / Pkg of 1
DuPont Liveo Pharma-65 Platinum-Cured Silicone Tubing, 1/4" ID x 7/16" OD; 50 Ft 5012203 Mfr # 4094222
COMPARE
1/4 7/16 3/32
$521.58CAD / Pkg of 1
DuPont Liveo Pharma-65 Platinum-Cured Silicone Tubing, 3/8" ID x 5/8" OD; 50 Ft 5012204 Mfr # 4008345
COMPARE
3/8 5/8 1/8
$404.65CAD / Pkg of 1
$875.19CAD / Pkg of 1
DuPont Liveo Pharma-65 Platinum-Cured Silicone Tubing, 1/2" ID x 3/4" OD; 50 Ft 5012205 Mfr # 4008346
COMPARE
1/2 3/4 1/8
$465.15CAD / Pkg of 1
$1,007.80CAD / Pkg of 1
DuPont Liveo Pharma-65 Platinum-Cured Silicone Tubing, 3/4" ID x 1" OD; 50 Ft 5012206 Mfr # 4008347
COMPARE
3/4 1 1/8
$679.70CAD / Pkg of 1
$1,468.97CAD / Pkg of 1
Showing 1- 9 of 9