Drierite Desiccants
Keeps samples dry and moisture free
  • Solves all types of drying problems
  • Can be regenerated repeatedly for reuse
Made from the naturally occurring mineral, gypsum (calcium sulfate), these desiccants maintain a dry atmosphere in storage spaces, vaults, commercial packages, and other enclosures. They also offer protection of hygroscopic materials or materials subject to mildew, corrosion, rust, or other deterioration caused by high humidity. Several varieties are available to solve all types of drying problems. Choose indicating or non-indicating desiccant.
Indicating desiccant is impregnated with cobalt chloride. It starts as a blue color when dry and changes to pink upon absorption of moisture. This allows you to know that dryness is being maintained and signals when the drying agent should be replaced.
Nonindicating desiccant features neutral pH, constant in volume, chemically inert except toward water, insoluble in organic liquids and refrigerants, non-disintegrating, non-wetting, non-toxic, non-corrosive, non-channeling, generally recognized as safe (GRAS) by the FDA, not regulated by OSHA, and is economical to use.
$170.18 - $707.23CAD / Case of 4
22 variations of this product are available.
See All Product Options Below
22 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Material Mesh Size
Availability Pricing
Drierite 23001 Indicating Desiccant, 8 Mesh; 1 lb Jar, 12/CS 0719305 Mfr # 23001
COMPARE
Blue to pink Indicating Desiccant - Anhydrous calcium sulfate 8
$456.75CAD / Carton of 12
Drierite 23005 Indicating Desiccant, 8 Mesh; 5 lb Jar, 4/CS 0719315 Mfr # 23005
COMPARE
Blue to pink Indicating Desiccant - Anhydrous calcium sulfate 8
$707.23CAD / Carton of 4
Drierite 13001 Nonindicating Desiccant, 8 Mesh; 1 lb Jar, 12/CS 0719320 Mfr # 13001
COMPARE
White Nonindicating Desiccant - Anhydrous calcium sulfate 8
$210.69CAD / Carton of 12
Drierite 13005 Nonindicating Desiccant, 8 Mesh; 5 lb Jar, 4/CS 0719325 Mfr # 13005
COMPARE
White Nonindicating Desiccant - Anhydrous calcium sulfate 8
$266.32CAD / Carton of 4
Drierite 11001 Nonindicating Desiccant,  4 Mesh; 1 lb Jar,  12/CS 0893409 Mfr # 11001
COMPARE
White Nonindicating Desiccant - Anhydrous calcium sulfate 4
$170.18CAD / Case of 12
Drierite 11005 Nonindicating Desiccant,  4 Mesh; 5 lb Jar, 4/CS 0893410 Mfr # 11005
COMPARE
White Nonindicating Desiccant - Anhydrous calcium sulfate 4
$210.69CAD / Case of 4
Drierite 11025 Nonindicating Desiccant,  4 Mesh; 25 lb Drum 0893411 Mfr # 11025
COMPARE
White Nonindicating Desiccant - Anhydrous calcium sulfate 4
$252.32CAD / Each
Drierite 12001 Nonindicating Desiccant,  6 Mesh; 1 lb Jar, 12/CS 0893412 Mfr # 12001
COMPARE
White Nonindicating Desiccant - Anhydrous calcium sulfate 6
$170.18CAD / Case of 12
Drierite 12005 Nonindicating Desiccant,  6 Mesh; 5 lb Jar, 4/CS 0893413 Mfr # 12005
COMPARE
White Nonindicating Desiccant - Anhydrous calcium sulfate 6
$210.69CAD / Case of 4
Drierite 12025 Nonindicating Desiccant,  6 Mesh; 25 lb Drum 0893414 Mfr # 12025
COMPARE
White Nonindicating Desiccant - Anhydrous calcium sulfate 6
$252.32CAD / Each
Drierite 14001 Nonindicating Desiccant, 10-20 Mesh; 1 lb Jar, 12/CS 0893415 Mfr # 14001
COMPARE
White Nonindicating Desiccant - Anhydrous calcium sulfate 10-20
$170.18CAD / Case of 12
Drierite 14005 Nonindicating Desiccant, 10-20 Mesh; 5 lb Jar, 4/CS 0893416 Mfr # 14005
COMPARE
White Nonindicating Desiccant - Anhydrous calcium sulfate 10-20
$210.69CAD / Case of 4
Drierite 14025 Nonindicating Desiccant,  10-20 Mesh; 25 lb Drum 0893417 Mfr # 14025
COMPARE
White Nonindicating Desiccant - Anhydrous calcium sulfate 10-20
$252.32CAD / Each
Drierite 21001 Indicating Desiccant, 4 Mesh; 1 lb Jar, 12/CS 0893418 Mfr # 21001
COMPARE
Blue to pink Indicating Desiccant - Anhydrous calcium sulfate 4
$342.19CAD / Case of 12
Drierite 21005 Indicating Desiccant, 4 Mesh; 5 lb Jar, 4/CS 0893419 Mfr # 21005
COMPARE
Blue to pink Indicating Desiccant - Anhydrous calcium sulfate 4
$570.94CAD / Case of 4
Drierite 21025 Indicating Desiccant, 4 Mesh; 25 lb Drum 0893420 Mfr # 21025
COMPARE
Blue to pink Indicating Desiccant - Anhydrous calcium sulfate 4
$651.98CAD / Each
Drierite 22001 Indicating Desiccant, 6 Mesh; 1 lb Jar, 12/CS 0893421 Mfr # 22001
COMPARE
Blue to pink Indicating Desiccant - Anhydrous calcium sulfate 6
$504.64CAD / Case of 12
Drierite 22005 Indicating Desiccant, 6 Mesh; 5 lb Jar, 4/CS 0893422 Mfr # 22005
COMPARE
Blue to pink Indicating Desiccant - Anhydrous calcium sulfate 6
$570.94CAD / Case of 4
Drierite 22025 Indicating Desiccant, 6 Mesh; 25 lb Drum 0893423 Mfr # 22025
COMPARE
Blue to pink Indicating Desiccant - Anhydrous calcium sulfate 6
$576.10CAD / Each
Drierite 24001 Indicating Desiccant, 10-20 Mesh; 1 lb Jar, 12/CS 0893424 Mfr # 24001
COMPARE
Blue to pink Indicating Desiccant - Anhydrous calcium sulfate 10-20
$349.93CAD / Case of 12
Showing 1 - 20 of 22