DILVAC® Portable Dry-Ice Maker
Quickly get dry ice when you need it
  • Compact, lightweight, and no electrical power is required
  • Makes a 2.2 lb (1 kg) solid block of dry ice in under 2 minutes
  • Dry ice blocks measure 3-1/8" x 4-3/8" x 7-1/8" (8 x 11 x 18 cm)
  • Connect to a 75 lb (34 kg) liquid CO2 cylinder at room temperature to yield 5 to 6 blocks
$2,733.14CAD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Dilvac® Portable Dry-Ice Maker
$2,733.14CAD / Each