DigiSense RTD Economical Cryogenic Probes
Designed For greater accuracies at low temperatures
  • Temperature ranges from -321°F - +300°F (-196°C to - +150°C)
  • Time Constant: 3 seconds or greater
  • Maximum error for probe assembly is ±1.1°C + 0.35% of reading below 300°F,. Alpha coefficient is 0.003850 Ù/Ù/°C (DIN IEC 751). Resistance at ice point (0°C) is 100 omega.
  • Custom designs or changes to this product are available upon request
Note: Time constant is the time required to reach 63.2% of the new reading. To determine how long it will take to reach 99% of the new reading multiply the time constant by five.
Note: the temperature range is for the probe tip only. The handle material may be affected if the probe is rated higher than the handle depending on exposure time, application, and handle material. ABS handles are rated to 220°F (104°C); PVC handles are rated to 200°F (93°C); Stainless Steel handles are rated at 450°F (232°C); Mini Connectors are rated to 392°F (200°C).
$134.08 - $140.71CAD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Connector Type Max Temperature (° F) Sheath Length (in)
Availability Pricing
COMPARE
Digi-Sense 3-pin 302 3
$137.03CAD / Each
COMPARE
Stripped Leads 302 3
$140.71CAD / Each
COMPARE
ANSI 3-blade 302 3
$140.71CAD / Each
COMPARE
Digi-Sense 3-pin 302 6
$134.08CAD / Each
COMPARE
Stripped Leads 302 6
$140.71CAD / Each
COMPARE
Digi-Sense 3-pin 302 12
$137.76CAD / Each