Digi-Sense Thermocouple Type R and Type S Probes
Type R and S Probes are Designed for Long Term Accurate Readings at Extremely High Temperatures
  • These probes feature ICONEL 600 sheaths w/ high purity MgO to prevent contamination of the platinum wires.
  • Type R and Type S are more accurate than other thermocouples at higher temperatures
  • Response Time: 2 seconds or greater
  • Each probe comes with a color coded ANSI mini connector: type R/S-green
  • Custom designs or changes to this product are available upon request
Note: the temperature range is for the probe tip only. The handle material may be affected if the probe is rated higher than the handle depending on exposure time, application, and handle material. ABS handles are rated to 220°F (104°C); PVC handles are rated to 200°F (93°C); Stainless Steel handles are rated at 450°F (232°C); Mini Connectors are rated to 392°F (200°C).
$1,003.38 - $1,025.48CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Probe Type
Availability Pricing
COMPARE
2100 R
$1,025.48CAD / Each
COMPARE
2100 S
$1,003.38CAD / Each