Digi-Sense Thermocouple General-Purpose Economical Probes
Hand held probes designed to be more compact and at a lower cost
  • Each probe comes with a 3/16” diameter shaft, 36” long FEP extension cable and handle
  • Available in grounded or ungrounded junctions.
  • Response Time: 2-4 seconds or greater
  • Each probe comes with a color coded ANSI mini connector: type J-black, type K-yellow, type T-blue, and type E-purple.
  • Custom designs or changes to this product are available upon request
Note: the temperature range is for the probe tip only. The handle material may be affected if the probe is rated higher than the handle depending on exposure time, application, and handle material. ABS handles are rated to 220°F (104°C); PVC handles are rated to 200°F (93°C); Stainless Steel handles are rated at 450°F (232°C); Mini Connectors are rated to 392°F (200°C).
$85.31 - $87.22CAD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Probe Type Sheath Length (in)
Availability Pricing
COMPARE
425 K 4
$87.08CAD / Each
COMPARE
425 K 6
$85.31CAD / Each
COMPARE
425 T 6
$85.31CAD / Each
COMPARE
425 J 6
$87.22CAD / Each
COMPARE
425 T 12
$87.22CAD / Each