Digi-Sense Thermocouple Bolt-On Surface Probes with Fiberglass Cable and Miniconnector
Bolt-On Probes with Fiberglass Insulated Cable Designed for Long Term Installation
  • Ideal for permanent installation inside ovens or other hot dry environments
  • Each Probe comes with a bolt on washer that is .62” OD and .28” ID for quick easy installation ideal for measuring machinery or mold temperatures
  • This probe is not recommended for damp or wet conditions
  • Response Time: 3 seconds or greater
  • Each probe comes with a color coded ANSI mini connector: type J-black, type K-yellow, type T-blue, and type E-purple.
  • Custom designs or changes to this product are available upon request
Note: the temperature range is for the probe tip only. The handle material may be affected if the probe is rated higher than the handle depending on exposure time, application, and handle material. ABS handles are rated to 220°F (104°C); PVC handles are rated to 200°F (93°C); Stainless Steel handles are rated at 450°F (232°C); Mini Connectors are rated to 392°F (200°C).
$97.10 - $102.84CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Probe Type
Availability Pricing
COMPARE
900 K
$102.84CAD / Each
COMPARE
900 J
$97.10CAD / Each
COMPARE
500 T
$101.37CAD / Each