Digi-Sense RTD Wire
Extend the measuring distance of your probes
  • 100-foot spool of RTD wire- cut to any lenght needed!
Three-strand RTD wire features excellent resistance to abrasion, chemicals, and moisture. Each silver-plated copper strad is color-coded with fluorinated ethylene propylene (FEP). The three starnds are twisted to increase flexibility and minimize electromagnetic noise, then jacketed in white FEP. The FEP insulation provides temperature resistance up to 400°F, and chemical resistance to solvents, acids, and oils. Additionally, the insulation protects the wire from thermal aging, while maintaining its strength and flexibility.
$279.21 - $542.21CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (ft) Max Temperature (° F) Wire Gauge (Ga)
Availability Pricing
COMPARE
100 900 24
$377.19CAD / Each
COMPARE
1000 400 20
$542.21CAD / Each
COMPARE
100 400 24
$279.21CAD / Each