Traceable® Temperature/Humidity Wi-Fi Data Loggers Compatible with TraceableLIVE® Cloud Service
Monitor environmental temperature and humidity from your mobile devices
  • Connect to your data via smartphone, tablet, or PC using TraceableLIVE wireless capability*
  • View current humidity and temperature conditions anywhere—cloud-based interface sends email, text, and push notifications
  • No software required—web, Android™, Apple® iOS data visibility
TraceableLIVE Wireless Technology allows multiple users to stay connected and monitor critical environments 24/7 wherever you go.
Remotely monitoring the temperature and humidity of laboratories, warehouses, and more has never been easier! Continuous data transmission using Wi-Fi provides a simple and reliable way to wirelessly monitor the environment.
Once setup is complete you can securely monitor parameters 24/7, control your alarms remotely, view your data logging history, run reports in real time, and give access to other team members. No local software is required to use the cloud-based data interface. Get mobile push notifications, emails, or texts for out-of-range conditions, connectivity interruptions, low-battery condition or if your device is due for calibration.
View current and Min/Max readings on the large tripe-line LCD. A convenient built-in bench stand makes it easy to read the temperature and humidity measurements. Select from model with a dongle or model with external probe and cable.
Traceable® Products come with an individually numbered Traceable® Certificate provided with each unit, assuring accuracy from our ISO/IEC 17025:2017 (1750.01) calibration laboratory, accredited by A2LA. The certificate indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMIs) that are signatories to the CIPM Mutual Recognition Agreement. Every Traceable® product undergoes an intense vetting process before it can carry the Traceable name, saving you both time and money by eliminating the need for separate calibration.
$576.10 - $609.25CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Input Type
Availability Pricing
Traceable Temperature/Humidity Wi-Fi Data Logger Compatible with TraceableLIVE® Cloud Service; 1 Dongle 1800029 Mfr # 6520TR
COMPARE
Dongle with Temp/Humidity Sensor
$576.10CAD / Each
Traceable Temperature/Humidity Wi-Fi Data Logger Compatible with TraceableLIVE® Cloud Service; 1 Probe with Cable 1800030 Mfr # 6521TR
COMPARE
Temp/Humdity Probe with Cable
$609.25CAD / Each