Digi-Sense Dual Thermocouple Connectors
Captive cover screw for easy assembly
  • Glass-filled nylon construction with molded barriers to prevent short circuit
  • Each connector comes in ANSI color coded material: type J-black, type K-yellow, and type T-blue
Each connector comes in ANSI color coded material: type J-black, type K-yellow, and type T-blue.
$29.47 - $162.07CAD / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Dual Thermocouple Connector, Standard, Male, Type-J; 1/ea
$39.63CAD / Each
COMPARE
Dual Thermocouple Connector, Standard, Female, Type-J; 1/ea
$32.71CAD / Each
COMPARE
Dual Thermocouple Connector, Standard, Male, Type-K; 1/ea
$32.56CAD / Each
COMPARE
Dual Thermocouple Connector, Standard, Male, Type-K; 5/pk
$137.61CAD / Pkg of 5
COMPARE
Dual Thermocouple Connector, Standard, Female, Type-K; 1/ea
$33.59CAD / Each
COMPARE
Dual Thermocouple Connector, Standard, Female, Type-K; 5/pk
$162.07CAD / Pkg of 5
COMPARE
Dual Thermocouple Connector, Standard, Female, Type-T; 1/ea
$38.01CAD / Each
COMPARE
Dual Thermocouple Connector, Mini, Female, Type-J; 1/ea
$29.47CAD / Each
COMPARE
Dual Thermocouple Connector, Mini, Male, Type-K; 1/ea
$31.24CAD / Each
COMPARE
Dual Thermocouple Connector, Mini, Male, Type-K; 5/pk
$107.85CAD / Pkg of 5
COMPARE
Dual Thermocouple Connector, Mini, Female, Type-K; 1/ea
$29.47CAD / Each
COMPARE
Dual Thermocouple Connector, Mini, Female, Type-K; 5/pk
$109.47CAD / Pkg of 5
Showing 1- 12 of 12