Digi-Sense Blue Spirit Verification Bottle Precision Accuracy Thermometers - Celsius Scale
Enclosed bottle ensures accuracy and is ideal for use in environmental chambers
  • An individually serial number is printed on the thermometer stem
  • Nonmercury blue spirit liquid
  • NIST- traceable statement of accuracy included
Accurately monitor temperatures in freezers, refrigerators, incubators, ovens, and other environments where stability is required. All thermometers are supplied in a glass bead filled unbreakable plastic bottle and can be removed from the bottle for recalibration. Use the magnetic attachment with pressure-sensitive backing to attach the bottle to a metal wall or door.
Thermometers feature permanently fused markings, blue spirit liquid, white background glass; are also individually serial numbered. Include an statement of accuracy and a triangular-style antiroll fitting.
$139.97 - $240.90CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° C) Min Temperature (° C)
Availability Pricing
COMPARE
-5 -25
$240.90CAD / Each
COMPARE
-5 -25
$208.48CAD / Each
COMPARE
10 -2
$139.97CAD / Each
COMPARE
30 15
$220.27CAD / Each