Digi-Sense Blue Spirit Laboratory Precision Accuracy Glass Thermometers, Celsius-Scale with Fractional-Degree
Scales are permanently fused on to the glass and won’t wear off
  • Nonmercury blue spirit liquid
  • Statement of accuracy
These thermometers are inexpensive and safe for use in laboratories, universities, food/beverage, environmental, wastewater, petrochemical, pharmaceutical and many other general-purpose applications. Fractional-degree thermometers offer higher precision. Choose from partial immersion or total immersion thermometers.
Thermometers feature permanently fused markings, blue spirit liquid, white background glass; are also individually serial numbered. Include a ring top and a statement of accuracy supplied by manufacturer.
$250.48 - $397.08CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Immersion Depth (mm) Max Temperature (° C) Min Temperature (° C)
Availability Pricing
COMPARE
76 101 -1
$397.08CAD / Each
COMPARE
610 101 -1
$250.48CAD / Each