Digi-Sense Blue Spirit Armored Glass Thermometers, Fahrenheit-scale
Armor shell absorbs shock and prevents thermometer damage
  • Nonmercury blue spirit liquid
  • Statement of accuracy included
Ideal for use in application where thermometers needs added protection from minor shocks. Nickel/plated brass armor shell protects these thermometers from accidental breakage during use and storage. The armor case has a screw cap with ring top and viewing window; the lower section that surrounds the bulb has small openings for circulation.
Thermometers feature permanently fused markings, blue spirit liquid, white background glass; are also individually serial numbered. Include statement of accuracy and a triangular-style antiroll fitting.
$73.67 - $78.83CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Min Temperature (° F) Overall Length (mm)
Availability Pricing
COMPARE
120 -40 300
$73.67CAD / Each
COMPARE
230 0 300
$73.67CAD / Each
COMPARE
300 0 300
$73.67CAD / Each
COMPARE
500 20 350
$78.83CAD / Each