Dibafit™ 1/4"-28 UNF(M) Gripper Fittings for 1/8"OD Tubing
One-piece grippers withstand pressures up to 1000 psi!
  • Permanently attached PTFE ferrules are ideal for repeated connect/disconnect
  • Inner PTFE sealing face is the only material in the fluid path
Dibafit gripper fitting system uses a one-piece ferrule that permanently attaches to the outside of semi-rigid tubing. The fitting can spin freely, preventing tubing from twisting. Select from polypropylene (PP), ETFE, or PEEK fittings. PP fittings are available in 10 different colors for easy identification. Mixed pack includes all 10 colors. ETFE and PEEK fittings provide excellent chemical resistance. PEEK fittings withstand higher temperatures and are available in compact or standard head.
$41.55 - $139.97CAD / Pkg of 10
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
Compareitem specifications
Color Fitting Material Description
Availability Pricing
Dibafit™ Gripper Fitting, 1/4"-28 UNF(M) flat bottom, 1/8" OD tubing, black; 10/pk 2193978 Mfr # 2200
COMPARE
Black Polypropylene Gripper Fitting, 1/4"-28 UNF(M) flat bottom, 1/8" OD tubing, black; 10/pk
$48.33CAD / Pkg of 10
Dibafit™ Gripper Fitting, 1/4"-28 UNF(M) flat bottom, 1/8" OD tubing, brown; 10/pk 2193979 Mfr # 2201
COMPARE
Brown Polypropylene Gripper Fitting, 1/4"-28 UNF(M) flat bottom, 1/8" OD tubing, brown; 10/pk
$41.55CAD / Pkg of 10
Dibafit™ Gripper Fitting, 1/4"-28 UNF(M) flat bottom, 1/8" OD tubing, red; 10/pk 2193980 Mfr # 002202
COMPARE
Red Polypropylene Gripper Fitting, 1/4"-28 UNF(M) flat bottom, 1/8" OD tubing, red; 10/pk
$48.33CAD / Pkg of 10
Dibafit™ Gripper Fitting, 1/4"-28 UNF(M) flat bottom, 1/8" OD tubing, orange; 10/pk 2193981 Mfr # 002203
COMPARE
Orange Polypropylene Gripper Fitting, 1/4"-28 UNF(M) flat bottom, 1/8" OD tubing, orange; 10/pk
$41.55CAD / Pkg of 10
Dibafit™ Gripper Fitting, 1/4"-28 UNF(M) flat bottom, 1/8" OD tubing, yellow; 10/pk 2193982 Mfr # 002204
COMPARE
Yellow Polypropylene Gripper Fitting, 1/4"-28 UNF(M) flat bottom, 1/8" OD tubing, yellow; 10/pk
$41.55CAD / Pkg of 10
Dibafit™ Gripper Fitting, 1/4"-28 UNF(M) flat bottom, 1/8" OD tubing, green; 10/pk 2193983 Mfr # 002205
COMPARE
Green Polypropylene Gripper Fitting, 1/4"-28 UNF(M) flat bottom, 1/8" OD tubing, green; 10/pk
$48.33CAD / Pkg of 10
Dibafit™ Gripper Fitting, 1/4"-28 UNF(M) flat bottom, 1/8" OD tubing, blue; 10/pk 2193984 Mfr # 002206
COMPARE
Blue Polypropylene Gripper Fitting, 1/4"-28 UNF(M) flat bottom, 1/8" OD tubing, blue; 10/pk
$41.55CAD / Pkg of 10
Dibafit™ Gripper Fitting, 1/4"-28 UNF(M) flat bottom, 1/8" OD tubing, violet; 10/pk 2193985 Mfr # 002207
COMPARE
Violet Polypropylene Gripper Fitting, 1/4"-28 UNF(M) flat bottom, 1/8" OD tubing, violet; 10/pk
$41.55CAD / Pkg of 10
Dibafit™ Gripper Fitting, 1/4"-28 UNF(M) flat bottom, 1/8" OD tubing, gray; 10/pk 2193986 Mfr # 002208
COMPARE
Gray Polypropylene Gripper Fitting, 1/4"-28 UNF(M) flat bottom, 1/8" OD tubing, gray; 10/pk
$48.33CAD / Pkg of 10
Dibafit™ Gripper Fitting, 1/4"-28 UNF(M) flat bottom, 1/8" OD tubing, white; 10/pk 2193987 Mfr # 002209
COMPARE
White Polypropylene Gripper Fitting, 1/4"-28 UNF(M) flat bottom, 1/8" OD tubing, white; 10/pk
$48.33CAD / Pkg of 10
Dibafit™ Gripper Fitting, 1/4"-28 UNF(M) flat bottom, 1/8" OD tubing, mixed; 10/pk 2193988 Mfr # 2210
COMPARE
Mixed Polypropylene Gripper Fitting, 1/4"-28 UNF(M) flat bottom, 1/8" OD tubing, mixed; 10/pk
$42.43CAD / Pkg of 10
Dibafit™ Compact Gripper Fitting, 1/4"-28 UNF(M) flat, 1/8" OD tubing, natural; 10/pk 2193989 Mfr # 002219
COMPARE
Natural PEEK Compact Gripper Fitting, 1/4"-28 UNF(M) flat, 1/8" OD tubing, natural; 10/pk
$112.27CAD / Pkg of 10
Dibafit™ Gripper Fitting, 1/4"-28 UNF(M) flat bottom, 1/8" OD tubing, natural; 10/pk 2193990 Mfr # 002222
COMPARE
Natural PEEK Gripper Fitting, 1/4"-28 UNF(M) flat bottom, 1/8" OD tubing, natural; 10/pk
$129.07CAD / Pkg of 10
Dibafit™ Gripper Fitting, 1/4"-28 UNF(M), 1/8" OD tubing, natural ETFE; 10/pk 2193992 Mfr # 2230
COMPARE
Natural ETFE Gripper Fitting, 1/4"-28 UNF(M), 1/8" OD tubing, natural ETFE; 10/pk
$139.97CAD / Pkg of 10
Showing 1- 14 of 14