Diba Omnifit® PTFE Cones (Ferrules) for Cap System Fittings and Valves
Select the size you need for a leaktight seal
  • Chemically resistant PTFE construction
Select PTFE cones when chemical resistance is important. The OD of the tubing should be matched to the inside diameter of the cone.
$36.10 - $66.16CAD / Pkg of 4
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 1) (mm) Description
Availability Pricing
Diba Omnifit® PTFE Cone (Ferrule) for Connector and Valve Caps, 1/16" (1.6 mm); 4/pk 2193947 Mfr # 1502
COMPARE
1.6 Omnifit® PTFE Cone (Ferrule) for Connector and Valve Caps, 1/16" (1.6 mm); 4/pk
$66.16CAD / Pkg of 4
Diba Omnifit® PTFE Cone (Ferrule) for Connector and Valve Caps, 1/8" (3.2 mm); 4/pk 2193948 Mfr # 001504
COMPARE
3.2 Omnifit® PTFE Cone (Ferrule) for Connector and Valve Caps, 1/8" (3.2 mm); 4/pk
$66.16CAD / Pkg of 4
Diba Omnifit® PTFE Cone (Ferrule) for Connector and Valve Caps, 1.5 mm; 4/pk 2193951 Mfr # 001508
COMPARE
1.5 Omnifit® PTFE Cone (Ferrule) for Connector and Valve Caps, 1.5 mm; 4/pk
$38.01CAD / Pkg of 4
Diba Omnifit® PTFE Cone (Ferrule) for Connector and Valve Caps, 2.0 mm; 4/pk 2193952 Mfr # 001509
COMPARE
2 Omnifit® PTFE Cone (Ferrule) for Connector and Valve Caps, 2.0 mm; 4/pk
$36.10CAD / Pkg of 4
Diba Omnifit® PTFE Cone (Ferrule) for Connector and Valve Caps, 3.0 mm; 4/pk 2193954 Mfr # 001511
COMPARE
3 Omnifit® PTFE Cone (Ferrule) for Connector and Valve Caps, 3.0 mm; 4/pk
$38.01CAD / Pkg of 4