Diba Omnifit® PTFE Cap System Tee Fittings and Valves
Connect various types of tubing with low dead volume
  • Use with 1 to 4 mm OD tubing
Quickly and easily connect semi-rigid tubing such as PTFE, ETFE, and FEP as well as rigid materials such as glass, PEEK, and stainless steel. Use the interchangeable components to create chemically inert fluid pathway connections and flat-bottom ports (using Gripper fittings). Fittings are autoclavable (remove caps first). Order colored caps quickly and easily color code multiple fluid lines.
$176.81 - $350.30CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Valve Material Orifice Dimensions (mm)
Availability Pricing
Diba Omnifit® Standard Connector for 1 to 4 mm OD Tubing, 3-way, tee; 1/ea 2193908 Mfr # 001010
COMPARE
PTFE 1.5
$176.81CAD / Each
Diba Omnifit® Standard Valve for 1 to 4 mm OD Tubing, 3-way, tee, single key; 1/ea 2193919 Mfr # 1109
COMPARE
PTFE 1.5
$289.89CAD / Each
Diba Omnifit® Small Bore Valve for 1 to 4 mm OD Tubing, 3-way, tee, single key; 1/ea 2194009 Mfr # 002422
COMPARE
PTFE 0.8
$350.30CAD / Each