Danger: Hazardous Chemicals Signs

Protect your workers by clearly displaying possible hazardous chemicals
These signs are intended to indicate potential, type, and degree of the hazard that may lead to accidental injury or property damage. Signs define specific hazards by conveying a clear, easily understood message. The sign headers and colors used in accident prevention safety signs are specified by OSHA and ANSI to designate the severity of the hazard. Each color has a specific role.
DANGER (red): indicates an imminently hazardous situation which, if not avoided, will result in death or serious injury.
$15.03 - $45.82CAD / Each

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Height (in) Material Width (in)
Availability Pricing
Accuform MCHL091VS Safety Sign, Danger - Hazardous Chemicals, 7" x 10", Adhesive Vinyl 4071163 Mfr # MCHL091VS
COMPARE
7 Vinyl 10
$15.03CAD / Each
Accuform MCHL091VP Safety Sign, Danger - Hazardous Chemicals, 7" x 10", Plastic 4071164 Mfr # MCHL091VP
COMPARE
7 Plastic 10
$17.68CAD / Each
Accuform MCHL092VA Safety Sign, Danger - Hazardous Chemicals, 10" x 14", Aluminum 4071168 Mfr # MCHL092VA
COMPARE
10 Aluminum 14
$45.82CAD / Each
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat